Skip to Content
Merck
All Photos(2)

Key Documents

P0076

Sigma-Aldrich

Anti-PINK1 antibody produced in rabbit

enhanced validation

~1.5 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-BRPK, Anti-PARK6, Anti-PTEN-induced putative kinase protein 1, Anti-Serine/threonine-protein kinase PINK1, mitochondrial precursor

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~60 kDa

species reactivity

mouse (predicted), human, rat (predicted)

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

concentration

~1.5 mg/mL

technique(s)

western blot: 1-2 μg/mL using HEK-293T cell lysate expressign human PINK1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PINK1(65018)
mouse ... Pink1(68943)
rat ... Pink1(298575)

Looking for similar products? Visit Product Comparison Guide

General description

PTEN induced putative kinase 1 (PINK1) is a serine/threonine kinase expressed in mitochondria. It contains an N-terminal mitochondrial targeting motif and a highly conserved kinase domain homologous to serine/threonine kinases of the Ca2+/calmodulin family.

Application

Anti-PINK1 antibody produced in rabbit has been used in immunoblotting.

Biochem/physiol Actions

PINK1 (PTEN induced putative kinase 1, also known as PARK6 and BRPK), has been identified as linked to the autosomal recessive form of familial Parkinson disease (PD). PINK1 localized to the mitochondria protects cells from stress-induced mitochondrial dysfunction. Overexpression of wild-type PINK1 has been found to protect neurons from stress-induced mitochondrial dysfunction and apoptosis. Genetic studies in drosophila indicate that PINK1 acts upstream of Parkin in a common pathway that influences mitochondrial morphology. PINK1 activity has been shown to protect primary neurons in mouse from the dopaminergic neurotoxin MPTP (1-methyl-4-phenylpyridine (MPP+)/1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine) both in vitro and in vivo. This protective activity requires PINK1 kinase activity, as a PINK1 G309D mutant linked to familial PD or a kinase dead mutant K219M are not protective. PINK1 has been shown to be cleaved and localized to the mitochondria, in response to enhanced proteasomal stress in vitro and correlates with increased expression of the processed PINK1 protein in PD brain.

Target description

PINK1 is a Ser/Thr kinase that has beenlocalized to the mitochondria, and thought to protectcells from stress-induced mitochondrial dysfunction.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PINK1/Parkin-mediated mitophagy is activated in cisplatin nephrotoxicity to protect against kidney injury
Wang Y, et al.
Cell Death & Disease, 9(11), 1113-1113 (2018)
Ying Wang et al.
Cell death & disease, 9(11), 1113-1113 (2018-11-06)
Cisplatin is a widely used chemotherapeutic drug with notorious toxicity in the kidneys, which involves mitochondrial dysfunction and damage in renal tubular cells. Mitophagy is a form of selective autophagy that removes damaged or dysfunctional mitochondria to maintain cellular homeostasis.
Agnese Gugliandolo et al.
Biomolecules, 13(8) (2023-08-26)
Parkinson's disease (PD) is a neurodegenerative disorder characterized by the loss of dopaminergic neurons in the substantia nigra and the accumulation of α-synuclein aggregates, known as Lewy bodies. It is known that mitochondria dysfunctions, including impaired localization, transport and mitophagy
Mutations in PTEN-induced putative kinase 1 associated with recessive parkinsonism have differential effects on protein stability
Beilina A, et al.
Proceedings of the National Academy of Sciences of the USA, 102(16), 5703-5708 (2005)
Ying Wang et al.
Redox biology, 38, 101767-101767 (2020-11-03)
Sepsis is the major cause of acute kidney injury (AKI) associated with high mortality rates. Mitochondrial dysfunction contributes to the pathophysiology of septic AKI. Mitophagy is an important mitochondrial quality control mechanism that selectively eliminates damaged mitochondria, but its role

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service