Skip to Content
Merck
All Photos(1)

Key Documents

P0027

Sigma-Aldrich

Pyrvinium pamoate salt hydrate

≥98% (HPLC)

Synonym(s):

6-(Dimethylamino)-2-[2-(2,5-dimethyl-1-phenyl-1H-pyrrol-3-yl)ethenyl]-1-methyl-4,4′-methylenebis[3-hydroxy-2-naphthalenecarboxylate] (2:1)-quinolinium, Alnoxin, Altolat, NSC 223622, PP, Pamovin, Vermitibier, Vipyrvinium embonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H28N3 · 0.5 C23H14O6 · xH2O
CAS Number:
Molecular Weight:
575.70 (anhydrous basis)
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Assay

≥98% (HPLC)

form

powder

color

red

solubility

DMSO: >10 mg/mL

storage temp.

room temp

SMILES string

O.CN(C)c1ccc2[n+](C)c(\C=C\c3cc(C)n(c3C)-c4ccccc4)ccc2c1.CN(C)c5ccc6[n+](C)c(\C=C\c7cc(C)n(c7C)-c8ccccc8)ccc6c5.Oc9c(cc%10ccccc%10c9Cc%11c(O)c(cc%12ccccc%11%12)C([O-])=O)C([O-])=O

InChI

1S/2C26H28N3.C23H16O6.H2O/c2*1-19-17-21(20(2)29(19)24-9-7-6-8-10-24)11-13-23-14-12-22-18-25(27(3)4)15-16-26(22)28(23)5;24-20-16(14-7-3-1-5-12(14)9-18(20)22(26)27)11-17-15-8-4-2-6-13(15)10-19(21(17)25)23(28)29;/h2*6-18H,1-5H3;1-10,24-25H,11H2,(H,26,27)(H,28,29);1H2/q2*+1;;/p-2

InChI key

FGNOZQNUEDSSTR-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Pyrvinium pamoate salt hydrate has been used as an activator of casein kinase 1α (CK1α) in terminal blast phase of chronic myeloid leukemia (BP-CML)[1] and human embryonic kidney (HEK293) cells.[2] It has also been used as an inhibitor of β-catenin in Wilms tumor[3]

Biochem/physiol Actions

Pyrvinium pamoate is a potent androgen receptor inhibitor.
Pyrvinium pamoate is a potent androgen receptor inhibitor. Androgen receptors (ARs) are nuclear hormone receptors/transcription factors that reside in the cytoplasm and activated by testosterone and dihydrotestosterone. AR inhibitors have potential therapeutic benefit in prostate cancer; competitive inhibitors and chemical castration methods have been discovered, but both therapies have undesirable side effects and/or resistance potential. A screen for non-competitive inhibitors was performed, resulting in the discovery of pyrvinium as an AR inhibitor. In comparison to competitive inhibitors, this compound does not bind to the ligand-binding domain of AR or block DNA occupancy by AR, but it inhibits AR-dependent gene expression via a distinct signaling mechanism. It is more potent than classical competitive AR antagonists and exhibits synergy with other AR inhibitors.

Features and Benefits

This compound is featured on the Nuclear Receptors (Steroids) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Casein kinase 1alpha-dependent feedback loop controls autophagy in RAS-driven cancers
Cheong JK, et al.
The Journal of Clinical Investigation, 125(4), 1401-1418 (2015)
Ajay K Verma et al.
Brain sciences, 9(7) (2019-07-13)
Autonomic reflex ascertains cardiovascular homeostasis during standing. Impaired autonomic reflex could lead to dizziness and falls while standing; this is prevalent in stroke survivors. Pulse rate variability (PRV) has been utilized in the literature in lieu of heart rate variability
Stephanie A Henson et al.
Nature communications, 8, 14682-14682 (2017-03-08)
Climate change is expected to modify ecological responses in the ocean, with the potential for important effects on the ecosystem services provided to humankind. Here we address the question of how rapidly multiple drivers of marine ecosystem change develop in
Pharmacologic inhibition of beta-catenin with pyrvinium inhibits murine and human models of wilms tumor
Polosukhina D, et al.
Oncology Research, 25(9), 1653-1664 (2017)
Pyrvinium selectively targets blast phase-chronic myeloid leukemia through inhibition of mitochondrial respiration
Xiang W, et al.
Testing, 6(32), 33769-33769 (2015)

Articles

We offers many products related to Nuclear Receptors (Steroids) for your research needs.

We offers many products related to Nuclear Receptors (Steroids) for your research needs.

We offers many products related to Nuclear Receptors (Steroids) for your research needs.

We offers many products related to Nuclear Receptors (Steroids) for your research needs.

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service