Skip to Content
Merck
All Photos(1)

Documents

NUC201

Sigma-Aldrich

Nuclei Isolation Kit: Nuclei PURE Prep

sufficient for 15 nuclei preparations (~1-10×107 cells or 1g of tissue per preparation)

Synonym(s):

Sucrose centrifugation nuclei isolation

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.32

usage

sufficient for 15 nuclei preparations (~1-10×107 cells or 1g of tissue per preparation)

Quality Level

packaging

pkg of 1 kit

storage condition

dry at room temperature

application(s)

cell analysis

foreign activity

nuclease and protease, free

shipped in

wet ice

storage temp.

2-8°C

Application

For preparation of pure nuclei and fragile nuclei from cell lines and solid tissues.

Biochem/physiol Actions

The protocol incorporates centrifugation through a dense sucrose cushion to protect nuclei and strip away cytoplasmic contaminants. High yield has been obtained from common cell lines (Jurkat, HFN7.1, COS7, HEK293 and MDCK) and tissues (spleen and liver). These preparations are suitable for many cell biology applications, e.g., as a source of nuclear components such as chromatin, genomic DNA, histones, and nuclear RNA/RNP, produces nuclei for in vitro apoptosis assays, and functional studies such as examination of the transcriptional status of cells.
The protocol incorporates centrifugation through a dense sucrose cushion to protect nuclei and strip away cytoplasmic contaminants. The sucrose concentration that is suitable for a particular cell type is determined empirically by the user. The sucrose concentrate and sucrose cushion buffer give the user flexibility to modify the density of the sucrose cushion as appropriate. High yield has been obtained from common cell lines (Jurkat, HFN7.1, COS7, HEK293 and MDCK) and tissues (spleen and liver). These preparations are suitable for many cell biology applications, e.g., as a source of nuclear components such as chromatin, genomic DNA, histones, and nuclear RNA/RNP, produces nuclei for in vitro apoptosis assays, and functional studies such as examination of the transcriptional status of cells.

Kit Components Only

Product No.
Description

  • Nuclei PURE Lysis Buffer 180 mL

related product

Product No.
Description
Pricing

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tyler G Ekins et al.
eLife, 9 (2020-11-06)
Type I lissencephaly is a neuronal migration disorder caused by haploinsuffiency of the PAFAH1B1 (mouse: Pafah1b1) gene and is characterized by brain malformation, developmental delays, and epilepsy. Here, we investigate the impact of Pafah1b1 mutation on the cellular migration, morphophysiology
Analysis of nuclear RNA, Chapter 14.
Robert E. Farrell, Jr., ed.
RNA Methodologies: A Laboratory Guide for Isolation and Characterization, 235-263 (1993)
Nuclei from rat liver: isolation method that combines purity with high yield.
G Blobel et al.
Science (New York, N.Y.), 154(3757), 1662-1665 (1966-12-30)
Identification of newly transcribed RNA.
Greenberg, M.E., and Bender, T.P., et al. et al.
Current Protocols in Molecular Biology, 4-4 (1987)
Baochen Shi et al.
BMC genomics, 10, 92-92 (2009-02-27)
A major goal of post-genomics research is the integrated analysis of genes, regulatory elements and the chromatin architecture on a genome-wide scale. Mapping DNase I hypersensitive sites within the nuclear chromatin is a powerful and well-established method of identifying regulatory

Articles

Centrifugation separates organelles based on size, shape, and density, facilitating subcellular fractionation across various samples.

Centrifugation separates organelles based on size, shape, and density, facilitating subcellular fractionation across various samples.

Centrifugation separates organelles based on size, shape, and density, facilitating subcellular fractionation across various samples.

Centrifugation separates organelles based on size, shape, and density, facilitating subcellular fractionation across various samples.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service