Skip to Content
Merck
All Photos(1)

Key Documents

NDEGLY

Sigma-Aldrich

Native Protein Deglycosylation Kit

Synonym(s):

Protein Deglycosylation Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

conjugate

(N-linked)

Quality Level

shipped in

wet ice

storage temp.

2-8°C

General description

All complex oligosaccharides can be reduced to the trimannosylchitobiose core by treatment of the glycoproteins with neuraminidase, β-galactosidase, and N-acetylglucosaminidase. Fucosidases may be required in some situations. The remaining trimannosylchitobiose core structures can be removed with endoglycosidase F3. Biantennary and triantennary structures can be immediately removed by endoglycosidases F2 and F3, respectively. Oligomannose and hybrid structures can be removed by Endoglycosidase F1.
For more information on each type of endoglycosidase, please refer to the Bulletin.
The Native Protein Deglycosylation is designed for the deglycoslylation of N-linked oligosaccharides from PNGase F-resistant native proteins. Endoglycosidases F1, F2, and F3 are less sensitive to protein conformation than PNGase F and are more suitable for removal of all classes of N-linked oligosaccharides without protein denaturation.

Application

Native Protein Deglycosylation Kit has been for N-deglycosylation of various enzymes such as laccase, tomato nuclease TBN1 and endo-β-1,3-glucanase.

Storage and Stability

The NDEGLY Kit ships on wet ice and storage at 2–8 °C is recommended. This kit may be used for at least 1 year when stored as indicated. Several days exposure to ambient temperatures will not reduce the activity of the enzymes in this kit.

Kit Components Only

Product No.
Description

  • Endoglycosidase F1 .3 U

  • Endoglycosidase F2 .1 U

  • Endoglycosidase F3 .1 U

  • Endoglycosidase F1 reaction buffer 200 μL

  • Endoglycosidase F2 & 3 reaction buffer 200 μL

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adela Rodríguez-Romero et al.
Acta crystallographica. Section D, Biological crystallography, 70(Pt 2), 329-341 (2014-02-18)
Endogenous glycosylated Hev b 2 (endo-β-1,3-glucanase) from Hevea brasiliensis is an important latex allergen that is recognized by IgE antibodies from patients who suffer from latex allergy. The carbohydrate moieties of Hev b 2 constitute a potentially important IgE-binding epitope
Amir E Zeituni et al.
Journal of bacteriology, 192(16), 4103-4110 (2010-06-22)
We recently reported that the oral mucosal pathogen Porphyromonas gingivalis, through its 67-kDa Mfa1 (minor) fimbria, targets the C-type lectin receptor DC-SIGN for invasion and persistence within human monocyte-derived dendritic cells (DCs). The DCs respond by inducing an immunosuppressive and
Junyi Ma et al.
Molecular pharmacology, 69(4), 1137-1145 (2006-01-10)
Opioid analgesics remain the choice for the treatment of moderate to severe pain. Recent research has established that the mu-opioid receptor is predominantly responsible for mediating many opioid actions, including analgesia and opioid tolerance. However, the function of delta-opioid receptors
N-glycosylation of tomato nuclease TBN1 produced in N. benthamiana and its effect on the enzyme activity
Podzimek T, et al.
Plant Science (2018)
Flavio Mena et al.
Neuroendocrinology, 91(1), 77-93 (2009-07-11)
We have previously shown that soluble factor(s) in conditioned media (CM) from the central and peripheral regions of the anterior pituitary (AP) gland of lactating rats promoted the in vitro dose-related release of prolactin (PRL) from pituitary glands of male

Articles

Find deglycosylation kits for your research. Review the different features of deglycosylation kits for glycoproteins, enzymatic proteins, and native proteins.

Find deglycosylation kits for your research. Review the different features of deglycosylation kits for glycoproteins, enzymatic proteins, and native proteins.

Find deglycosylation kits for your research. Review the different features of deglycosylation kits for glycoproteins, enzymatic proteins, and native proteins.

Find deglycosylation kits for your research. Review the different features of deglycosylation kits for glycoproteins, enzymatic proteins, and native proteins.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service