Skip to Content
Merck
All Photos(1)

Documents

N7265

Sigma-Aldrich

Nitrate Reductase (NAD[P]H) from Aspergillus niger

lyophilized powder, ≥300 units/g solid

Synonym(s):

NAD(P)H:Nitrate oxidoreductase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

form

lyophilized powder

Quality Level

specific activity

≥300 units/g solid

shipped in

wet ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Nitrate reductase (NAD[P]H) is expressed in variety of organisms including bacteria, fungi, algae, and higher plants which have capacity to use nitrate as their sole source of nitrogen.Experimental studies show that nitrate reductase (NAD[P]H) is 180,000Daltons.

Biochem/physiol Actions

Nitrate Reductase (NAD[P]H) catalyzes the reduction of nitrate to nitrite. In addition, it also catalyzes the reduction of NADPH-cytochrome c, FADH2-nitrate. Nitrate reductase also has reduced methyl viologen-nitrate reductase activities.

Caution

light and moisture sensitive

Unit Definition

One unit will reduce 1.0 μmole of nitrate per min in the presence of β-NADPH at pH 7.5 at 25 °C.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P C Allen et al.
Poultry science, 76(6), 822-827 (1997-06-01)
Two experiments were conducted to determine effects of diets containing n-3 fatty acids (n-3FA) from whole as well as ground flaxseed on the performance of broilers during coccidia infections. Diets were fed from 1 d of age through 3 wk
G Unden et al.
Molecular microbiology, 25(2), 205-210 (1997-07-01)
The FNR (fumarate and nitrate reductase regulation) protein of Escherichia coli is an oxygen-responsive transcriptional regulator required for the switch from aerobic to anaerobic metabolism. In the absence of oxygen, FNR changes from the inactive to the active state. The
W H Campbell
Cellular and molecular life sciences : CMLS, 58(2), 194-204 (2001-04-06)
Pyridine nucleotide-dependent nitrate reductases (NRs; EC 1.6.6.1-3) are molybdenum-containing enzymes found in eukaryotic organisms which assimilate nitrate. NR is a homodimer with an approximately 100 kDa polypeptide which folds into stable domains housing each of the enzyme's redox cofactors--FAD, heme-Fe
R L Warner et al.
Plant physiology, 91, 947-953 (1989-01-01)
Barley (Hordeum vulgare L.) has NADH-specific and NAD(P)H-bispecific nitrate reductase isozymes. Four isogenic lines with different nitrate reductase isozyme combinations were used to determine the role of NADH and NAD(P)H nitrate reductases on nitrate transport and assimilation in barley seedlings.
Nilesh Dharajiya et al.
The Journal of allergy and clinical immunology, 119(3), 646-653 (2007-03-06)
Ragweed extract (RWE) contains NADPH oxidases that induce oxidative stress in the airways independent of adaptive immunity (signal 1) and augment antigen (signal 2)-induced allergic airway inflammation. To test whether inhibiting signal 1 by administering antioxidants inhibits allergic airway inflammation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service