Skip to Content
Merck
All Photos(2)

Key Documents

N5632

Sigma-Aldrich

Nile Blue A

Dye content ≥75 %, Powder

Synonym(s):

Basic Blue 12, Nile blue sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2C20H20N3O · SO4
CAS Number:
Molecular Weight:
732.85
Colour Index Number:
51180
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Product Name

Nile Blue A, Dye content ≥75 %

form

powder

composition

Dye content, ≥75%

mp

>300 °C (dec.) (lit.)

solubility

H2O: 1 mg/mL, blue

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[O-]S([O-])(=O)=O.CC\[N+](CC)=C1/C=CC2=Nc3c(OC2=C1)cc(N)c4ccccc34.CC\[N+](CC)=C5/C=CC6=Nc7c(OC6=C5)cc(N)c8ccccc78

InChI

1S/2C20H19N3O.H2O4S/c2*1-3-23(4-2)13-9-10-17-18(11-13)24-19-12-16(21)14-7-5-6-8-15(14)20(19)22-17;1-5(2,3)4/h2*5-12,21H,3-4H2,1-2H3;(H2,1,2,3,4)

InChI key

QIRDPEPUXNCOLD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nile Blue A is aminodiethylaminonaphthophenoxazonium sulfate employed in biology and histology. It is a highly fluorescent, and photostable organic dye used to dye clothing, and is for use in lipid histochemistry.

Application

Nile blue A is suitable for staining and discriminating between melanins and lipofuscins in paraffin sections of animal tissue, and in a histochemical method for lipids in frozen sections.

Biochem/physiol Actions

Nile blue A stains acidic components, such as phospholipids, nucleic and fatty acids by imparting a dark blue color, while neutral lipids, namely triglycerides, are stained pink, or red.

Features and Benefits

Used in the preparation of an amperometric glucose sensor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Segmental origin and migration of neural crest cells in the hindbrain region of the chick embryo.
Lumsden A
Development, 113, 1281-1281 (1991)
Fgfr1 regulates patterning of the pharyngeal region.
Trokovic N
Genes & Development, 17, 141-141 (2003)
Biochemical, histochemical and ultrastructural characterization of Centrolobium robustum (Fabaceae) seeds.
Pinzon-Torres JA
Hoehnea, 36, 149-149 (2009)
Rost FWD
Fluorescence Microscopy (1992)
Zhiping Cui et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 83(1), 1-7 (2011-09-29)
A dual-wavelength overlapping resonance Rayleigh scattering (DWO-RRS) method was developed to detect chondroitin sulfate (CS) with nile blue sulfate (NBS). At pH 3.0-4.0 Britton-Robinson (BR) buffer medium, CS interacted with NBS to form an ion-association complex. As a result, the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service