Skip to Content
Merck
All Photos(1)

Key Documents

N3254

Sigma-Aldrich

4-Nitrophenyl phosphate di(tris) salt

phosphatase substrate, ≥97% (enzymatic), powder

Synonym(s):

4-Nitrophenyl phosphate bis(tris) salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6 H4 N O6 P)( C4 H12 N O3)2
CAS Number:
Molecular Weight:
461.36
Beilstein:
1578142
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

Product Name

4-Nitrophenyl phosphate di(tris) salt, phosphatase substrate

Quality Level

Assay

≥97% (enzymatic)

form

powder

solubility

water: 50 mg/mL, clear, colorless to light yellow

storage temp.

−20°C

SMILES string

NC(CO)(CO)CO.NC(CO)(CO)CO.OP(O)(=O)Oc1ccc(cc1)[N+]([O-])=O

InChI

1S/C6H6NO6P.2C4H11NO3/c8-7(9)5-1-3-6(4-2-5)13-14(10,11)12;2*5-4(1-6,2-7)3-8/h1-4H,(H2,10,11,12);2*6-8H,1-3,5H2

InChI key

XXAXKCWOTRABOW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Nitrophenyl phosphate di(tris) salt has been used:
  • as a substrate to quantify tartrate-resistant acid phosphatase (TRAP) activity in cell lysates
  • to determine phosphatase activity using native polyacrylamide gel electrophoresis (PAGE)
  • for alkaline phosphatase in enzyme-linked immunosorbent assay (ELISA)

Biochem/physiol Actions

p-Nitrophenyl Phosphate (pNPP) is a preferred substrate for alkaline phosphatase in enzyme-linked immunosorbent assay (ELISA) procedures, because of its high sensitivity. This substrate produces a soluble yellow end-product and can be read spectrophotometrically at 405 nm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julieta B Carrillo et al.
PloS one, 13(1), e0191621-e0191621 (2018-01-24)
Ostreococcus tauri, the smallest free-living (non-symbiotic) eukaryote yet described, is a unicellular green alga of the Prasinophyceae family. It has a very simple cellular organization and presents a unique starch granule and chloroplast. However, its starch metabolism exhibits a complexity
Enzyme-linked Immunosorbent Assay (ELISA): From A to Z null
B Le Gars Santoni et al.
Bioactive materials, 10, 222-235 (2021-12-14)
β-Tricalcium Phosphate (β-TCP), one of the most used bone graft substitutes, may contain up to 5 wt% foreign phase according to standards. Typical foreign phases include β-calcium pyrophosphate (β-CPP) and hydroxyapatite (HA). Currently, the effect of small amounts of impurities on
Mary E Wood et al.
Vaccine, 36(50), 7737-7743 (2018-11-12)
Chronic wasting disease (CWD) is a fatal prion disease affecting multiple cervid species. Effective management tools for this disease, particularly in free-ranging populations, are currently limited. We evaluated a novel CWD vaccine in elk (Cervus canadensis) naturally exposed to CWD
B Le Gars Santoni et al.
Bioactive materials, 10, 222-235 (2021-12-14)
β-Tricalcium Phosphate (β-TCP), one of the most used bone graft substitutes, may contain up to 5 wt% foreign phase according to standards. Typical foreign phases include β-calcium pyrophosphate (β-CPP) and hydroxyapatite (HA). Currently, the effect of small amounts of impurities on

Articles

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service