Skip to Content
Merck
All Photos(1)

Key Documents

MSST0057

Sigma-Aldrich

SILuProt IL8, Interleukin-8 human

recombinant, expressed in HEK 293 cells, SIL MS Protein Standard, 13C and 15N-labeled

Synonym(s):

C-X-C motif chemokine 8 chemokine (C-X-C motif), Emoctakin, Granulocyte chemotactic protein 1 (GCP-1), IL-8, Monocyte-derived neutrophil, Monocyte-derived neutrophil-activating peptide (MONAP), Neutrophil-activating protein 1 (NAP-1), Protein 3-10C, T-cell chemotactic factor, chemotactic factor (MDNCF), ligand 8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23201100
NACRES:
NA.32

recombinant

expressed in HEK 293 cells

Quality Level

Assay

≥95% (SDS-PAGE)

form

lyophilized powder

potency

≥98% (Heavy amino acids incorporation efficiency by MS)

suitability

suitable for mass spectrometry (standard)

UniProt accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

human ... CXCL8(3576)

Related Categories

General description

SILuProt IL8 is a recombinant, stable isotope-labeled human CXCL8 which incorporates [13C6, 15N4]-Arginine and [13C6, 15N2]-Lysine. Expressed in human 293 cells, it is designed to be used as an internal standard for bioanalysis of CXCL8 in mass-spectrometry. SILuProt IL8 is a mixture of the 3 main CXCL8 isoforms with the molecular weights, amino acid number and relative abundance as described in Table 1 of the product′s datasheet.

Biochem/physiol Actions

Interleukin-8 (IL-8) is a member of the CXC chemokine subfamily and is produced by blood cells and many types of tissues. The measurement of IL-8 in voided urinary samples may have utility for urine-based detection of bladder cancer. Urinary IL-8 was a strong biomarker of stress under intensive and prolonged demands, both acutely and over time. IL-8 and cathepsin B levels were significantly elevated in melanoma patients, and more importantly, the combination of IL-8 and cathepsin B were also studied as a prognosis marker for melanoma mortality.

Sequence

EGAVLPRSAKELRCQCIKTYSKPFHPKFIKELRVIESGPHCANTEIIVKLSDGRELCLDPKENWVQRVVEKFLKRAENS

Physical form

Supplied as a lyophilized powder containing phosphate buffered saline.

Legal Information

This product is licensed under U.S. Patent No. 7,396,688 and foreign counterparts from E. I. du Pont de Nemours and Company. The purchase of this product conveys to the buyer the nontransferable right to use the purchased amount of the product for research and development only, including services for a third party for consideration. The buyer cannot sell or otherwise transfer this product, its components or materials made using this product or its components to a third party. Information about licenses for excluded uses is available from: E. I. du Pont de Nemours and Company; Attn: Associate Director, Commercial Development; DuPont Experimental Station E268; 200 Powdermill Rd.; Wilmington, DE 19803; 1-877-881-9787 (voice), 1-302-695-1437 (fax), [email protected].
SILu is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service