Skip to Content
Merck
All Photos(1)

Documents

MHS128

Sigma-Aldrich

Mayer′s Hematoxylin Solution

Synonym(s):

(6aS,11bR)-7,11b-Dihydroindeno[2,1-c]chromene-3,4,6a,9,10(6H)-pentol, Delafield′s hematoxylin, Progressive stains

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41116124
PubChem Substance ID:
NACRES:
NA.47

form

solution

Quality Level

shelf life

Expiry date on the label.

IVD

for in vitro diagnostic use

concentration

1 g/L

pH

2.4 (25 °C)

application(s)

hematology
histology

storage temp.

room temp

SMILES string

Oc1cc2C[C@@]3(O)COc4c(O)c(O)ccc4[C@H]3c2cc1O

InChI

1S/C16H14O6/c17-10-2-1-8-13-9-4-12(19)11(18)3-7(9)5-16(13,21)6-22-15(8)14(10)20/h1-4,13,17-21H,5-6H2/t13-,16+/m0/s1

InChI key

WZUVPPKBWHMQCE-XJKSGUPXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Used as a counterstain for procedures such as immunohistochemistry or laser microdissection.

Application

General purpose nuclear stain, progressive type. Used with hematoxylin and eosin staining.

Other Notes

1 g/L certified hematoxylin

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stephany Orjuela et al.
BMC cancer, 20(1), 290-290 (2020-04-08)
Identifying molecular differences between primary and metastatic colorectal cancers-now possible with the aid of omics technologies-can improve our understanding of the biological mechanisms of cancer progression and facilitate the discovery of novel treatments for late-stage cancer. We compared the DNA
Shun-Jie Chen et al.
Oncology reports, 37(2), 1148-1158 (2016-12-14)
Acute kidney injury (AKI) is a disease where kidney function is lost almost instantaneously; it can develop very rapidly over few hours to maximum of few days. Despite the advent of technology, the clinical management against this disease is very
Adam Smith et al.
Molecular cancer therapeutics, 8(6), 1666-1675 (2009-06-11)
Although viridins like wortmannin (Wm) have long been examined as anticancer agents, their ability to self-activate has only recently been recognized. Here, we describe the cytostatic effects of a self-activating viridin (SAV), which is an inactive, polymeric prodrug. SAV self-activates
Li-Gen Lin et al.
Journal of medicinal chemistry, 51(15), 4419-4429 (2008-07-10)
Protein tyrosine kinase (PTK) inhibitors represent emerging therapeutics for cancer chemoprevention. In our study, hematoxylin (26) was identified as one of the most remarkable c-Src inhibitors in an orthogonal compound-mixing library (32200 compounds) by using an ELISA-based automated high-throughput screening
Hideki Ishii et al.
Bioorganic & medicinal chemistry letters, 22(3), 1469-1474 (2012-01-17)
SAR studies for the exploration a novel class of anti-human immunodeficiency virus type 1 (HIV-1) agents based on the hematoxylin structure (1) are described. The systematic deoxygenations of 1 including asymmetric synthesis were conducted to obtain a compound showing high

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service