MAK157
Renin Assay Kit
sufficient for 100 fluorometric tests
Synonym(s):
Renin Enzyme Detection Kit
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
12161503
NACRES:
NA.84
Recommended Products
usage
sufficient for 100 fluorometric tests
detection method
fluorometric
relevant disease(s)
cardiovascular diseases
storage temp.
−20°C
Gene Information
human ... REN(5972)
rat ... REN(24715)
Related Categories
General description
Renin, also known as angiotensinogenase, is an aspartyl protease that cleaves angiotensinogen to form angiotensin I, the initial and rate-limiting step of the renin-angiotensin system (RAS). This system plays a critical role in the regulation of blood pressure and fluid balance. Increased renin activity can result in hypertension due to increased vasoconstriction and fluid retention. Inhibitors of renin can be used as a treatment for hypertension.
Application
Renin Assay Kit has been used to determine the activity of renin.
Suitability
The Renin Assay Kit provides a convenient and simple method for screening renin activity in a variety of samples or for screening renin inhibitors.
Principle
Renin activity is determined using a proprietary fluorescence resonance energy transfer (FRET) peptide containing a fluorescence quencher. Cleavage of the peptide by renin results in the production of a fluorescent product (λex = 540/λem = 590 nm). This assay is ~50-fold more sensitive than the EDANS/DABCYL-based assay.
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Árpád Kovács et al.
American journal of physiology. Heart and circulatory physiology, 310(11), H1671-H1682 (2016-04-10)
Hypertension (HTN) is a major risk factor for heart failure. We investigated the influence of HTN on cardiac contraction and relaxation in transgenic renin overexpressing rats (carrying mouse Ren-2 renin gene, mRen2, n = 6). Blood pressure (BP) was measured.
Aliskiren decreases oxidative stress and angiogenic markers in retinal pigment epithelium cells.
Sim?o S, et al.
Angiogenesis, 20(1), 175-181 (2017)
The hormone FGF21 stimulates water drinking in response to ketogenic diet and alcohol.
Song P, et al.
Cell Metabolism, 27(6), 1338-1347 (2018)
Caroline G Shimoura et al.
Journal of applied physiology (Bethesda, Md. : 1985), 128(5), 1329-1337 (2020-04-03)
Acute intermittent hypoxia (AIH) triggers sympathetic long-term facilitation (sLTF), a progressive increase in sympathetic nerve activity (SNA) linked to central AT1 receptor (AT1R) activation by circulating angiotensin II (ANG II). Here, we investigated AIH activation of the peripheral renin-angiotensin system
Yoshio T, et al.
Handbook of Hormones: Comparative Endocrinology for Basic and Clinical Research (2015)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service