MAK118
Invertase Activity Assay Kit
sufficient for 100 colorimetric or fluorometric tests
Synonym(s):
β-fructofuranosidase Assay Kit
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
usage
sufficient for 100 colorimetric or fluorometric tests
application(s)
cosmetics
environmental
food and beverages
detection method
colorimetric
fluorometric
storage temp.
−20°C
General description
Invertase (β-fructofuranosidase) is an enzyme that catalyzes the hydrolysis of sucrose to fructose and glucose. Invertase is produced by certain plants, microorganisms, and honey bees.
Application
Invertase Assay Kit has been used for screening inhibition of recombinant invertase inhibitor protein from sugarcane.
Features and Benefits
Compatible with high-throughput handling systems.
Suitability
Suitable for the detection of invertase activity in biological and environmental samples and for the screening of invertase inhibitors.
Principle
In this kit, invertase activity is determined by a coupled enzyme assay in which invertase cleaves sucrose to glucose and fructose, resulting in a colorimetric (570 nm)/fluorometric (λex = 535/λem = 587 nm) product, proportional to the invertase activity present. One unit of Invertase is the amount of enzyme that will catalyze the formation of 1.0 μmole of glucose per minute at pH 4.5 under the assay conditions.
Storage Class Code
10 - Combustible liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Frontiers in plant science, 9, 1887-1887 (2019-01-09)
Hexokinases is a family of proteins that is found in all eukaryotes. Hexokinases play key roles in the primary carbon metabolism, where they catalyze the phosphorylation of glucose and fructose, but they have also been shown to be involved in
Animals : an open access journal from MDPI, 10(5) (2020-05-14)
The purpose of this study was to examine the effects of dietary inclusion of two additives at the final concentration of 0.5% bilberry (E1) and 1% walnut (E2) leaves powder in the basal diet on digestive health of hens. A
Journal of experimental botany, 67(11), 3303-3312 (2016-04-17)
Invertases are a widespread group of enzymes that catalyse the conversion of sucrose into fructose and glucose. Plants invertases and their substrates are essential factors that play an active role in primary metabolism and in cellular differentiation and by these
Nature communications, 10(1), 4680-4680 (2019-10-17)
Date palms (Phoenix dactylifera) are an important fruit crop of arid regions of the Middle East and North Africa. Despite its importance, few genomic resources exist for date palms, hampering evolutionary genomic studies of this perennial species. Here we report
Bio-protocol, 8(10), e2856-e2856 (2018-05-20)
Invertase can catalyze the hydrolysis of sucrose, and is widely distributed in cells of cyanobacteria and plants. Being responsible for the first step for sucrose metabolism, invertase plays important physiological roles and its enzymatic activity is frequently needed to be
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service