Skip to Content
Merck
All Photos(1)

Key Documents

M9019

Sigma-Aldrich

IgG3, Lambda from murine myeloma

clone Y5606, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Mouse IgG3-λ

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

clone

Y5606, monoclonal

form

buffered aqueous solution

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

IgG (Immunoglobulin G) are the most common Immunoglobulin′s isotype in blood, lymph, cerebrospinal and peritoneal fluids and serve as key players in the humoral immune response. IgG include four subclasses (IgG1, IgG2, IgG3, and IgG4), consist of a variable Fab fragment (which includes the antibody recognition site) and a conserved Fc fragment.

Application

IgG3, λ from murine myeloma is suitable as isotype control in immunohistochemistry of formalin-fixed, paraffin-embedded sections and in FACS.
IgG3, Lambda from murine myeloma antibody has been used in Fluorescent activated cell sorting (FACS).

Biochem/physiol Actions

IgG antibody subtype is the most abundant of serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defense of the neonate against infections.

Physical form

Solution in 0.02 M Tris buffered saline, pH 8.0, containing 0.02% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nurlaely Mida Rachmawati et al.
International immunology, 25(2), 117-128 (2012-10-19)
Stimulation of Toll-like receptor 4 (TLR4) induces not only innate but also adaptive immune responses, and has been suggested to exert adjuvant effects. Additional to such positive effects, pre-stimulation of TLR4 induces endotoxin tolerance where animals are unresponsive to subsequent
Nicholas R Forsyth et al.
Cloning and stem cells, 8(1), 16-23 (2006-03-31)
Human embryonic stem cells (hESC) have great potential in regenerative medicine, provided that culture systems are established that maintain genomic integrity. Here we describe a comparison of the effects of culture in either physiologic oxygen (2%) or room oxygen (21%)
Systematic analysis of ATG13 domain requirements for autophagy induction
Wallot-Hieke N, et al.
Autophagy, 14(5), 743-763 (2018)
Tracy Goodpaster et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 56(4), 347-358 (2007-12-12)
Fibroblasts are critical for tissue homeostasis, and their inappropriate proliferation and activation can result in common and debilitating conditions including fibrosis and cancer. We currently have a poor understanding of the mechanisms that control the growth and activation of fibroblasts
Gestur Vidarsson et al.
Frontiers in immunology, 5, 520-520 (2014-11-05)
Of the five immunoglobulin isotypes, immunoglobulin G (IgG) is most abundant in human serum. The four subclasses, IgG1, IgG2, IgG3, and IgG4, which are highly conserved, differ in their constant region, particularly in their hinges and upper CH2 domains. These

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service