M9015
Malachite Green oxalate salt
Dye content, ≥90%, certified by the Biological Stain Commission, powder or crystals
Synonym(s):
N,N,N′,N′-Tetramethyl-4,4′-diaminotriphenylcarbenium oxalate, Basic Green 4
About This Item
Recommended Products
Product Name
Malachite Green oxalate salt, certified by the Biological Stain Commission
grade
certified by the Biological Stain Commission
form
powder or crystals
composition
Dye content, ≥90%
technique(s)
microbe id | staining: suitable
color
faint green to very dark green
mp
144-150 °C
solubility
H2O: 1 mg/mL, blue to very deep blue
application(s)
diagnostic assay manufacturing
hematology
histology
storage temp.
room temp
SMILES string
C[N+](C)=C1C=CC(C=C1)=C(C2=CC=C(N(C)C)C=C2)C3=CC=CC=C3.[O-]C(C(O)=O)=O
InChI
1S/C23H25N2.C2H2O4/c1-24(2)21-14-10-19(11-15-21)23(18-8-6-5-7-9-18)20-12-16-22(17-13-20)25(3)4;3-1(4)2(5)6/h5-17H,1-4H3;(H,3,4)(H,5,6)/q+1;/p-1
InChI key
REZFCYOVFCJTDG-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
General description
Application
- in the preparation of Alexander staining solution for cell viability test
- in malachite green-ammonium molybdate assay for determining the adenosine triphosphatase (ATPase) activity of binding immunoglobulin protein (BiP)
- as a hydrophobic ion, to study the label-free molecular transport in the living cell membranes of Escherichia coli in real time by second-harmonic generation (SHG) method
Suitability
Other Notes
927.00 g/mol - the mass of 2 mol malachite green + 2 mol monohydrogen oxalate + 1 mol dihydrogen oxalate, or
463.50 g/mol - the mass of 1 mol malachite green + 1 mol monohydrogen oxalate + 0.5 mol dihydrogen oxalate
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Repr. 2
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service