M7523
Anti-Myosin (Skeletal) antibody produced in rabbit

whole antiserum
About This Item
Recommended Products
biological source
rabbit
conjugate
unconjugated
antibody form
whole antiserum
antibody product type
primary antibodies
clone
polyclonal
contains
15 mM sodium azide
species reactivity
human
enhanced validation
independent
Learn more about Antibody Enhanced Validation
technique(s)
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:20 using human or animal skeletal muscle
indirect immunofluorescence: 1:20 using human or animal sletetal muscle
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... MYH13(8735) , MYH3(4621) , MYL1(4632)
General description
Specificity
Immunogen
Application
- immunohistochemistry
- immunostaining
- western blotting
- immunofluorescence
- immunolocalization
- immunocytochemistry
Immunocytochemistry (1 paper)
Western Blotting (1 paper)
Quality
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service