Skip to Content
Merck
All Photos(3)

Key Documents

M6500

Sigma-Aldrich

Cysteamine hydrochloride

powder or crystals, ≥98% (titration)

Synonym(s):

β-Mercapto­ethylamine hydrochloride, 2-Amino­ethane­thiol hydrochloride, 2-Mercapto­ethyl­amine hydrochloride, Decarboxy­cysteine hydrochloride, Thio­ethanol­amine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2CH2NH2 · HCl
CAS Number:
Molecular Weight:
113.61
Beilstein:
3590083
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

Product Name

Cysteamine hydrochloride, ≥98% (titration)

biological source

synthetic (organic)

Assay

≥98% (titration)

form

powder or crystals

mp

67-71 °C

solubility

water: 50 mg/mL, clear, colorless to very faintly yellow

storage temp.

2-8°C

SMILES string

Cl[H].NCCS

InChI

1S/C2H7NS.ClH/c3-1-2-4;/h4H,1-3H2;1H

InChI key

OGMADIBCHLQMIP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Cysteamine derived from cysteine degradation is the simplest stable aminothiol. Cysteamine is used in the biosynthesis of coenzyme A (CoA). Cysteamine may be useful in studies on oxidative stress and glycation (transglycating agent). Cysteamine is a substrate useful to study the specificity and kinetics of cysteamine (2-aminoethanethiol) dioxygenase(s) (ADO).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anne-Sophie Racine et al.
Molecular brain, 14(1), 130-130 (2021-08-26)
Somatostatin-expressing interneurons (SOM-INs) are a major subpopulation of GABAergic cells in CA1 hippocampus that receive excitation from pyramidal cells (PCs), and, in turn, provide feedback inhibition onto PC dendrites. Excitatory synapses onto SOM-INs show a Hebbian long-term potentiation (LTP) mediated
Matthieu Lagardère et al.
Scientific reports, 10(1), 19954-19954 (2020-11-19)
Fluorescence live-cell and super-resolution microscopy methods have considerably advanced our understanding of the dynamics and mesoscale organization of macro-molecular complexes that drive cellular functions. However, different imaging techniques can provide quite disparate information about protein motion and organization, owing to
Ca Leisinger et al.
Animal reproduction science, 150(1-2), 70-75 (2014-09-28)
The present study utilized a 2×2×2 factorial design examining age (old vs. young), follicle size (≥2mm vs. <2mm) and media supplementation (with or without fetal bovine serum [FBS]) to determine factors that might affect in vitro maturation of alpaca oocytes.
Duncan E Scott et al.
Cell chemical biology, 28(6), 835-847 (2021-03-05)
BRCA2 controls RAD51 recombinase during homologous DNA recombination (HDR) through eight evolutionarily conserved BRC repeats, which individually engage RAD51 via the motif Phe-x-x-Ala. Using structure-guided molecular design, templated on a monomeric thermostable chimera between human RAD51 and archaeal RadA, we
Edward P Harvey et al.
Cell chemical biology, 27(6), 647-656 (2020-05-16)
The BCL-2 family is composed of anti- and pro-apoptotic members that respectively protect or disrupt mitochondrial integrity. Anti-apoptotic overexpression can promote oncogenesis by trapping the BCL-2 homology 3 (BH3) "killer domains" of pro-apoptotic proteins in a surface groove, blocking apoptosis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service