Skip to Content
Merck
All Photos(3)

Key Documents

M5757

Sigma-Aldrich

Maleic acid sodium salt

~99%

Synonym(s):

cis-Butenedioic acid, Toxilic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C4H3O4Na
CAS Number:
Molecular Weight:
138.05
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.77

Assay

~99%

SMILES string

OC(/C=C\C([O-])=O)=O.[Na+]

InChI

1S/C4H4O4.Na/c5-3(6)1-2-4(7)8;/h1-2H,(H,5,6)(H,7,8);/q;+1/p-1/b2-1-;

InChI key

VRVKOZSIJXBAJG-ODZAUARKSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Biochem/physiol Actions

Maleic acid is a component of cosmetics, fragrances and has industrial applications. It is also a potent nephrotoxin and is implicated to induce glycosuria, aminoaciduria and Fanconi syndrome.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mei-Ling Hou et al.
Molecules (Basel, Switzerland), 21(3), 367-367 (2016-03-22)
Maleic acid has been shown to be used as a food adulterant in the production of modified starch by the Taiwan Food and Drug Administration. Due to the potential toxicity of maleic acid to the kidneys, this study aimed to
Pierangelo Gobbo et al.
Langmuir : the ACS journal of surfaces and colloids, 28(33), 12357-12363 (2012-08-14)
Improved methodology to prepare maleimide-functionalized, water-soluble, small (<3 nm) gold nanoparticles using a retro-Diels-Alder strategy that we developed for similar organic-soluble AuNP's is described. Importantly, our results suggest that a recent paper by Zhu, Waengler, Lennox, and Schirrmacher describing a
P Pandi et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 88, 77-81 (2012-01-10)
Synthesis and growth of a novel organic nonlinear optical (NLO) crystal of 4-aminopyridinium maleate (4APM) in larger size by the slow evaporation solution growth technique are reported. Single crystal and powder X-ray diffraction analyses reveal that 4APM crystallizes in monoclinic
Nidambur Vasudev Ballal et al.
Journal of endodontics, 37(10), 1402-1405 (2011-09-20)
The elimination of microorganisms from the root canal system necessitates the use of combination of irrigating solutions to enhance their antimicrobial property. The combination of irrigants and their interaction sometimes could be detrimental to the outcome of the root canal
Marek Drozd et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 89, 243-251 (2012-01-24)
On the basis of experimental literature data the theoretical studies for guanidinium and maleic acid complex with using DFT method are performed. In these studies the experimental X-ray data for two different forms of investigated crystal were used. During the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service