Skip to Content
Merck
All Photos(1)

Key Documents

M5391

Sigma-Aldrich

(±)-Metoprolol (+)-tartrate salt

≥98% (titration), powder

Synonym(s):

(±)1-(Isopropylamino)-3-[p-(β-methoxyethyl)phenoxy]-2-propanol (+)-tartrate salt, Lopressor

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C15H25NO3)2 · C4H6O6
CAS Number:
Molecular Weight:
684.81
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (titration)

form

powder

color

white to off-white

solubility

H2O: 50 mg/mL

originator

Novartis

SMILES string

O[C@H]([C@@H](O)C(O)=O)C(O)=O.COCCc1ccc(OCC(O)CNC(C)C)cc1.COCCc2ccc(OCC(O)CNC(C)C)cc2

InChI

1S/2C15H25NO3.C4H6O6/c2*1-12(2)16-10-14(17)11-19-15-6-4-13(5-7-15)8-9-18-3;5-1(3(7)8)2(6)4(9)10/h2*4-7,12,14,16-17H,8-11H2,1-3H3;1-2,5-6H,(H,7,8)(H,9,10)/t;;1-,2-/m..1/s1

InChI key

YGULWPYYGQCFMP-CEAXSRTFSA-N

Gene Information

human ... ADRB1(153)

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Metoprolol has antihypertensive and anti-ischemic properties. It is used to treat arrhythmias.

Application

(±)-Metoprolol (+)-tartrate salt has been used:
  • to modulate the immunological parameters in various studies
  • to study its effects onCa2+-calmodulin-dependent protein kinase II (CaMKII) activity in human and experimental heart failure (HF)
  • to examine the contribution of the adrenergic receptors to the production of norepinephrine (NE)-mediated O2-

Biochem/physiol Actions

β1-adrenoceptor antagonist.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound was developed by Novartis. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Other Notes

Shelf-life of the powder is at least 4 years.

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Metoprolol
Koch-Weser J
The New England Journal of Medicine, 301(13), 698-703 (1979)
The acute modulation of norepinephrine on immune responses and genes expressions via adrenergic receptors in the giant freshwater prawn, Macrobrachium rosenbergii
Chang CC, et al.
Fish & Shellfish Immunology, 46(2), 459-467 (2015)
Octopamine enhances the immune responses of freshwater giant prawn, Macrobrachium rosenbergii, via octopamine receptors
Kuo HW and Cheng W
Developmental and Comparative Immunology, 81, 19-32 (2018)
Tyramine's modulation of immune resistance functions in Litopenaeus vannamei and its signal pathway
Kuo HW, et al.
Developmental and Comparative Immunology (2019)
Mitochondrial superoxide signaling contributes to norepinephrine-mediated T-lymphocyte cytokine profiles
Case AJ, et al.
PLoS ONE, 11(10), e0164609-e0164609 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service