Skip to Content
Merck
All Photos(1)

Key Documents

M4879

Sigma-Aldrich

3-O-Methyl-D-glucopyranose

≥98% (GC)

Synonym(s):

3-O-Methyl-α-D-glucopyranose, 3-O-Methylglucose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H14O6
CAS Number:
Molecular Weight:
194.18
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Assay

≥98% (GC)

form

powder

optical activity

[α]25/D 55 to 57 °, c = 1% (w/v) in water + trace NH4OH

technique(s)

gas chromatography (GC): suitable

mp

167-169 °C (lit.)

solubility

water: 50 mg/mL, clear to slightly hazy, colorless

SMILES string

CO[C@@H]1[C@@H](O)[C@@H](O)O[C@H](CO)[C@H]1O

InChI

1S/C7H14O6/c1-12-6-4(9)3(2-8)13-7(11)5(6)10/h3-11H,2H2,1H3/t3-,4-,5-,6+,7+/m1/s1

InChI key

SCBBSJMAPKXHAH-OVHBTUCOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-O-Methyl-D-glucopyranose is a passive carrier-mediated transported glucose analogue.

Application

Varying concentrations of 3-O-methyl-D-glucopyranose were used to measure responses of jejunal and ileal short circuit currents to determine maximal transport capacity and carrier affinity in a study to assess intestinal glucose transport in rats with diabetes mellitus. It has also been used in a study to investigate nonmetabolizable glucose analogues and ornithine decarboxylase expression in LLC-PK1 cells.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R N Fedorak et al.
Canadian journal of physiology and pharmacology, 69(8), 1143-1148 (1991-08-01)
Chronic diabetes enhances intestinal absorption of glucose and induces hyperphagia. We examined the enhanced intestinal absorption of glucose in ad libitum-fed rats with streptozocin-induced diabetes mellitus and compared these results with those obtained from pair-fed diabetic animals. Maximal transport capacity
Nam Q Nguyen et al.
Obesity surgery, 26(1), 77-84 (2015-05-20)
The aim was to determine the effects of fat and protein preloads on pouch emptying (PE), caecal arrival time (CAT), glucose absorption, blood glucose (BSL), gut hormones, haemodynamics and gastrointestinal (GI) symptoms in subjects who had undergone Roux-en-Y gastric bypass
D W Lundgren et al.
The American journal of physiology, 259(4 Pt 1), C647-C653 (1990-10-01)
This report examines the effect of nonmetabolizable glucose analogues on ornithine decarboxylase (ODC) activity in LLC-PK1 cells. The addition of Na(+)-dependent cotransported glucose analogues, 1-O-methyl-alpha-D-glucopyranoside (alpha-MDG) and 1-O-methyl-beta-D-glucopyranoside, to Earle's balanced salt solution minus glucose (EBSS-G) increased ODC activity five-
Anna E Di Bartolomeo et al.
Critical care (London, England), 16(5), R167-R167 (2012-09-19)
Studies in the critically ill that evaluate intragastric and post-pyloric delivery of nutrient have yielded conflicting data. A limitation of these studies is that the influence in the route of feeding on glucose absorption and glycaemia has not been determined.
Asaf Miller et al.
Critical care and resuscitation : journal of the Australasian Academy of Critical Care Medicine, 19(1), 37-42 (2017-02-22)
To evaluate the effect of exogenous glucagonlike peptide-1 (GLP-1) on small intestinal glucose absorption and blood glucose concentrations during critical illness. A prospective, blinded, placebo-controlled, cross-over, randomised trial in a mixed medical-surgical adult intensive care unit, with 12 mechanically ventilated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service