Skip to Content
Merck
All Photos(3)

Key Documents

M3885

Sigma-Aldrich

MES sodium salt

≥99% (titration)

Synonym(s):

2-(N-Morpholino)ethanesulfonic acid sodium salt, 4-Morpholineethanesulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12NNaO4S
CAS Number:
Molecular Weight:
217.22
Beilstein:
3765682
EC Number:
MDL number:
UNSPSC Code:
12161700
eCl@ss:
32129211
PubChem Substance ID:
NACRES:
NA.25

Assay

≥99% (titration)

form

crystalline powder

impurities

≤8% water content (Karl Fischer)

useful pH range

5.5-6.7

pKa 

6.1

solubility

water: 0.5 g/mL, clear, colorless

application(s)

diagnostic assay manufacturing

SMILES string

[Na+].[O-]S(=O)(=O)CCN1CCOCC1

InChI

1S/C6H13NO4S.Na/c8-12(9,10)6-3-7-1-4-11-5-2-7;/h1-6H2,(H,8,9,10);/q;+1/p-1

InChI key

IRHWMYKYLWNHTL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

MES sodium salt is commonly employed as buffer. Useful pH range for MES sodium salt is 5.5 to 6.7.

Application

MES sodium salt is suitable for cell culture and diagnostic assay manufacturing. It was used to prepare buffer to study the expression profiling by microarrays. It may be used as buffer to investigate the enzymatic degradation of DNA to nucleosides by P1 nuclease.

Other Notes

Easily compare specifications for MES sodium salt products with the MES sodium salt specification table.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Precise measurement of the G+ C content of deoxyribonucleic acid by high-performance liquid chromatography.
Mesbah M, et al.
International J. Systemic Bact., 39(2), 159-167 (1989)
Bianca Santos Henriques et al.
Frontiers in physiology, 11, 509310-509310 (2021-02-02)
Rhodnius prolixus is one important vector for the parasite Trypanosoma cruzi in Latin America, where Chagas disease is a significant health issue. Although R. prolixus is a model for investigations of vector-parasite interaction and transmission, not much has been done
Tsuyoshi Shirai et al.
Journal of molecular biology, 321(5), 879-889 (2002-09-11)
The crystal structure of congerin II, a galectin family lectin from conger eel, was determined at 1.45A resolution. The previously determined structure of its isoform, congerin I, had revealed a fold evolution via strand swap; however, the structure of congerin
Takamitsu Miyafusa et al.
Acta crystallographica. Section F, Structural biology and crystallization communications, 64(Pt 6), 512-515 (2008-06-10)
Capsular polysaccharides (CPs) are important virulence factors of Staphylococcus aureus. The biosynthesis of type 5 and type 8 CPs (CP5 and CP8), which are produced by most clinical isolates of S. aureus, is catalyzed by 16 CP-assembling proteins. One of
Theresa E Bankston et al.
Journal of chromatography. A, 1217(14), 2123-2131 (2010-03-03)
Hydroxyapatite (HAP) columns, widely used for chromatographic separation of proteins and other biomolecules because of their unique selectivity and ability to resolve complex mixtures, exhibit limited stability at acidic conditions requiring careful control of pH. Even with buffered solutions, however

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service