Skip to Content
Merck
All Photos(1)

Key Documents

M3164

Sigma-Aldrich

14-Methylhexadecanoic acid

Synonym(s):

(+)-14-methyl palmitic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H34O2
CAS Number:
Molecular Weight:
270.45
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic (organic)

Quality Level

Assay

≥96% (GC)

form

powder

functional group

carboxylic acid

lipid type

saturated FAs

shipped in

ambient

storage temp.

2-8°C

SMILES string

CCC(C)CCCCCCCCCCCCC(O)=O

InChI

1S/C17H34O2/c1-3-16(2)14-12-10-8-6-4-5-7-9-11-13-15-17(18)19/h16H,3-15H2,1-2H3,(H,18,19)

InChI key

FXUKWLSZZHVEJD-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ramona Jenske et al.
Chemistry and physics of lipids, 154(1), 26-32 (2008-04-15)
Differential scanning calorimetry (DSC) has been applied to study the effect of free hydroxylated and methyl-branched fatty acids on the physico-chemical properties of lipid membranes. First, the impact of free hydroxy fatty acids (HFAs) on dimyristoylphosphatidylcholine (DMPC) model membranes was
R L Wolff et al.
Lipids, 32(9), 971-973 (1997-10-06)
14-Methylhexadecanoic (14-MHD) acid has been identified in a sample of pine seed oil (Pinus contorta) by gas-liquid chromatography-mass spectrometry of its picolinyl ester derivative. Its identification (through its equivalent chain length) and its distribution in four conifer families have been
Simone Hauff et al.
Journal of chromatography. A, 1217(10), 1683-1687 (2010-02-02)
Gas chromatography in combination with electron ionization mass spectrometry (GC/EI-MS) was used to determine the fatty acids of a membrane lipid from Bacillus megaterium. Special attention was put on the structure and absolute configuration of a monoenoic fatty acid previously
Z Tuhácková et al.
Neoplasma, 32(3), 323-334 (1985-01-01)
Labeled 14-methylhexadecanoic acid was administered to normal rats, animals bearing the Walker 256 tumor at various stages of its growth and to tumor-resistant rats and its distribution in lipids (free fatty acids, triglycerides, phospholipids and cholesteryl esters) was studied during
Simone Hauff et al.
Lipids, 45(4), 357-365 (2010-03-12)
Anteiso fatty acids (aFA) are substituted with a methyl group on the antepenultimate carbon of the straight acyl chain. This feature leads to a stereogenic center. The 12-methyltetradecanoic acid (a15:0) and the 14-methylhexadecanoic acid (a17:0) are the most common aFA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service