Skip to Content
Merck
All Photos(1)

Key Documents

L090000

Sigma-Aldrich

Dichloromethane

Synonym(s):

Methylene chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH2Cl2
CAS Number:
Molecular Weight:
84.93
Beilstein:
1730800
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.21
bp:
39.8-40 °C (lit.)
vapor pressure:
24.45 psi ( 55 °C)
6.83 psi ( 20 °C)

vapor density

2.9 (vs air)

vapor pressure

24.45 psi ( 55 °C)
6.83 psi ( 20 °C)

type

for DNA synthesis

product line

Proligo Reagents

form

liquid

autoignition temp.

1223 °F

contains

amylene as stabilizer

expl. lim.

22 %

storage condition

dry at room temperature

impurities

≤50 ppm water content (Karl Fischer)

color

clear

refractive index

n/D 1.421-1.425
n20/D 1.424 (lit.)

bp

39.8-40 °C (lit.)

mp

−95 °C (lit.)

density

1.325 g/mL at 25 °C (lit.)

SMILES string

ClCCl

InChI

1S/CH2Cl2/c2-1-3/h1H2

InChI key

YMWUJEATGCHHMB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

In the solid-phase synthesis of oligonucleotides, liquid reagents are used in each step of the synthesis cycle. Overall synthesis performance, and therefore total product yield and purity of the crude oligonucleotide, is highly dependent on the chemical purity of the monomers and the supporting liquid reagents and other solvents like ethylacetate, dichloromethane, and dimethylformadine .Dichloromethane is an ordinary organic solvent frequently utilized in peptide synthesis. Under normal circumstances, DCM maintains highly stable and will not cause significant side reactions in peptide synthesis. It is a chlorinated organic solvent that is susceptible to degradation with time. This can be suppressed by adding amylene as a stabilizer.Dichloromethane has been tested as a solvent medium for the dipyridine-chromium(VI) oxide. Solubility was reported to be 12.5g/100ml. The role of the quantity of TiO2 loading on activated carbon support employed in the photodecomposition of dichloromethane has been investigated.

Application

Dichloromethane may be used as a solvent for the preparation of stock solutions of carotenoids. It may be employed as a solvent in the synthesis of polylactide-based (PLA) thin films. IT is also suitable as a solvent for the oxidation of primary and secondary alcohols using dipyridine-chromium(VI) oxide.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Khalef Lefsih et al.
Phytochemistry, 154, 47-55 (2018-07-15)
This work aims to fill the gap in the present knowledge about the structure of pectin from Opuntia ficus-indica. The water-soluble pectin (WSP) fraction, extracted with the Microwave Assisted Extraction (MAE), was further deproteinated (dWSP) and analyzed through several biophysical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service