Skip to Content
Merck
All Photos(1)

Key Documents

L0636

Sigma-Aldrich

Lectin from Triticum vulgaris (wheat)

suitable for cell culture, BioReagent, lyophilized powder

Synonym(s):

WGA, Wheat germ agglutinin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

sterility

aseptically processed

Quality Level

product line

BioReagent

form

lyophilized powder

technique(s)

cell culture | mammalian: suitable

impurities

salt, essentially free

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Lectin from Triticum vulgaris (wheat) is a dimeric glycoprotein of 36 kDa. It belongs to the family of chitin-binding proteins. Glycosylated precursor of lectin is synthesized in endoplasmic reticulum.

Application

Lectin from Triticum vulgaris (wheat) has been used:
  • to conjugate with N-hydroxysuccinimide (NHS) dye
  • to inhibit nuclear import
  • to inhibit the nuclear pore of human cervical carcinoma HeLa and SiHa cells

Biochem/physiol Actions

Lectin from Triticum vulgaris (wheat) possessing four binding sites is specific for N-acetyl-d-glucosamine and N-acetyl-d-neuraminic acid (sialic acid) residues. Lectins also function as a fungicide. They also possess anti-nutritive effects.
WGA is not blood group specific but has an affinity for N-acetyl-β-D-glucosaminyl residues and N-acetyl-β-D-glucosamine oligomers. WGA contains no protein-bound carbohydrate.

Other Notes

Highly purified

Preparation Note

Conjugates are prepared from affinity purified lectin.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effects of wheat germ agglutinin on human gastrointestinal epithelium: insights from an experimental model of immune/epithelial cell interaction
Dalla P, et al.
Toxicology and Applied Pharmacology, 237, 146-153 (2009)
Super-multiplex vibrational imaging
Wei L, et al.
Nature, 544, 465-465 (2017)
Pengfei Zhang et al.
Chemical science, 13(43), 12760-12768 (2022-12-16)
Exosome analysis is a promising tool for clinical and biological research applications. However, detection and biomarker quantification of exosomes is technically challenging because they are small and highly heterogeneous. Here, we report an optical approach for imaging exosomes and quantifying
Plant lectins: occurrence, biochemistry, functions and applications
Rudiger H and Gabius HJ
Glycoconjugate Journal, 18, 589-613 (2001)
Jonathan Welch et al.
Regulatory toxicology and pharmacology : RTP, 125, 105022-105022 (2021-08-02)
The aim of the study was to use multiple in vitro assays to assess the effects of a model irritant, sodium dodecyl sulphate (SDS) (≤10 mM (0.29 %, w/v)), on an in vitro model of the airway, MucilAir™. The use of MucilAir™

Articles

Find the right lectin for your research with our lectin selection guide, organized by lectin source/species, carbohydrate specificity, blood group specificity, and more.

Find the right lectin for your research with our lectin selection guide, organized by lectin source/species, carbohydrate specificity, blood group specificity, and more.

Find the right lectin for your research with our lectin selection guide, organized by lectin source/species, carbohydrate specificity, blood group specificity, and more.

Find the right lectin for your research with our lectin selection guide, organized by lectin source/species, carbohydrate specificity, blood group specificity, and more.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service