K3129
Kao and Michayluk Vitamin Solution
100 ×, liquid, suitable for plant cell culture
Synonym(s):
Nutrient Solution
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352207
NACRES:
NA.72
Recommended Products
sterility
sterile-filtered
form
liquid
concentration
100 ×
technique(s)
cell culture | plant: suitable
color
yellow
pH
4.0-6.5
application(s)
agriculture
shipped in
dry ice
storage temp.
−20°C
General description
Kao and Michayluk Vitamin Solution K3129 is a 100-fold concentrate of the vitamins contained in Kao and Michayluk media. It may be used in combination with Kao and Michayluk basal salts to reconstituted Kao and Michayluk media.
Application
Kao and Michayluk Vitamin Solution has been used:
- as a media supplement to induce callus induction
- in liquid marine salts medium, to test the requirements for strain MOLA115 culture
- as a component in LeMaster culture
Reconstitution
Use at a concentration of 10 mL per liter of prepared medium to achieve the proper final concentration.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Irnov Irnov et al.
PLoS genetics, 13(8), e1006978-e1006978 (2017-08-23)
To achieve robust replication, bacteria must integrate cellular metabolism and cell wall growth. While these two processes have been well characterized, the nature and extent of cross-regulation between them is not well understood. Here, using classical genetics, CRISPRi, metabolomics, transcriptomics
Clémence Bonnot et al.
The New phytologist, 223(2), 993-1008 (2019-04-05)
ROOT HAIR DEFECTIVE SIX-LIKE (RSL) genes control the development of structures from single cells at the surface of embryophytes (land plants) such as rhizoids and root hairs. RSL proteins constitute a subclass (VIIIc) of the basic helix-loop-helix (bHLH) class VIII
Michael R Davey et al.
Methods in molecular biology (Clifton, N.J.), 318, 201-210 (2006-05-06)
The family Passifloraceae contains many species exploited in the food, pharmaceutical, and ornamental plant industries. The routine culture of isolated protoplasts (naked cells) followed by reproducible plant regeneration, is crucial to the genetic improvement of Passiflora spp. by somatic cell
Crystal structure of butyrate kinase 2 from Thermotoga maritima, a member of the ASKHA superfamily of phosphotransferases
Diao J and Hasson MS
Journal of Bacteriology, 191(8), 2521-2529 (2009)
Pleionea mediterranea gen. nov., sp. nov., a gammaproteobacterium isolated from coastal seawater
Fagervold SK, et al.
International Journal of Systematic and Evolutionary Microbiology, 63(7), 2700-2705 (2013)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service