K2000
α-Ketoglutaric acid potassium salt
≥98% (enzymatic)
Synonym(s):
2-Oxopentanedioic acid potassium salt
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C5H5O5K
CAS Number:
Molecular Weight:
184.19
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32
Recommended Products
Assay
≥98% (enzymatic)
form
powder
solubility
water: 100 mg/mL, clear, colorless to faintly yellow
storage temp.
2-8°C
SMILES string
[K+].OC(=O)CCC(=O)C([O-])=O
InChI
1S/C5H6O5.K/c6-3(5(9)10)1-2-4(7)8;/h1-2H2,(H,7,8)(H,9,10);/q;+1/p-1
InChI key
XTCZBVVKDHLWKU-UHFFFAOYSA-M
Related Categories
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Li-Li Chen et al.
Journal of molecular modeling, 19(4), 1919-1927 (2013-01-23)
Bacillus subtilis natto is widely used in industry to produce natto, a traditional and popular Japanese soybean food. However, during its secondary fermentation, high amounts of ammonia are released to give a negative influence on the flavor of natto. Glutamate
Piotr Dobrowolski et al.
Nutrition (Burbank, Los Angeles County, Calif.), 29(3), 556-561 (2012-12-12)
Proton-pump inhibitors, such as omeprazole, are widely used in the prevention and treatment of gastroesophageal diseases. However, an association between proton-pump inhibitors and the increased risk of bone fractures has been observed, especially in patients treated for extended periods. Conversely
Michael E Jung et al.
The Journal of organic chemistry, 77(23), 11002-11005 (2012-11-21)
Oxidative cleavage of cycloalkene-1-carboxylates, made from the corresponding carboxylic acids, and subsequent oxidation of the resulting ketoaldehyde afforded the important 1-monoesters of 2-ketoalkanedioic acids. Thus ozonolysis of octyl cyclobutene-1-carboxylate followed by sodium chlorite oxidation afforded the 1-monooctyl 2-ketoglutarate. This is
Vivian R Moure et al.
Journal of bacteriology, 195(2), 279-286 (2012-11-13)
Fe protein (dinitrogenase reductase) activity is reversibly inactivated by dinitrogenase reductase ADP-ribosyltransferase (DraT) in response to an increase in the ammonium concentration or a decrease in cellular energy in Azospirillum brasilense, Rhodospirillum rubrum, and Rhodobacter capsulatus. The ADP-ribosyl is removed
Sylwia Magiera et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 919-920, 20-29 (2013-02-16)
A simple, accurate and sensitive liquid chromatography tandem mass spectrometry (LC-MS/MS) method was developed and validated for the quantitation of α-ketoglutaric acid (α-KG), L-carnitine (L-CAR) and acetyl-L-carnitine (acetyl-L-CAR) in human urine as potential biomarkers of cardiovascular disease. The separation was
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service