Skip to Content
Merck
All Photos(1)

Key Documents

INHIB1

Sigma-Aldrich

Protease Inhibitor Kit

powder, for the inhibition of serine, cysteine, calpains and metalloproteinases, for custom cocktail preparation

Synonym(s):

Protease inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.77
form:
powder
solubility:
water: soluble
storage temp.:
−20°C

Product Name

Protease Inhibitor Panel, for custom cocktail preparation

form

powder

solubility

water: soluble

storage temp.

−20°C

Related Categories

General description

As powders, all reagents can be stored at 0 °C. Those reagents designated for room temperature and 2-8 °C storage do not require storage at 0 °C, but will not be adversely affected when stored at 0 °C. Allow all powders to warm to room temperature before opening. Store tightly sealed and protect from moisture.

Application

Create your own broad-spectrum protease inhibitor cocktails or screen your extracts for proteolytic activity. Panel includes inhibitors for serine, cysteine, calpains and metalloproteinases. Components also include the economical alternatives NEM, EAC and trypsin inhibitor.
Stock solutions of each inhibitor should be prepared first, prior to creating a cocktail. Mixtures of some inhibitor stock solutions may result in precipitation due to interactions between inhibitors and the mixing of solvents. In most cases further dilution will aid solubility.

Other Notes

All kit components are also available separately.

Kit Components Only

Product No.
Description

  • E-64 5 mg

  • AEBSF 25 mg

  • Antipain 5 mg

  • Aprotinin 5 mg

  • Benzamidine HCl 5 g

  • Bestatin 5 mg

  • Chymostatin 5 mg

  • EDTA 50 g

  • ε-aminocaproic acid 25 g

  • N-Ethylmaleimide 5 g

  • Leupeptin 5 mg

  • Pepstatin A 5 mg

  • Phosphoramidon 5 mg

  • Trypsin inhibitor 100 mg

See All (14)

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1A - Skin Sens. 1 - STOT RE 2 Inhalation

Target Organs

Respiratory Tract

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Transcriptomic analysis of pea plant responses to chitooligosaccharides' treatment revealed stimulation of mitogen-activated protein kinase cascade.
Kozyulina, et al.
Frontiers in Plant Science, 14, 1092013-1092013 (2023)
Bart Bryant et al.
PloS one, 6(11), e25502-e25502 (2011-11-30)
Autophagy plays a pivotal role by allowing cells to recycle cellular components under conditions of stress, starvation, development and cancer. In this work, we have demonstrated that programmed autophagy in the mosquito fat body plays a critical role in maintaining
Camila Boniche-Alfaro et al.
Frontiers in immunology, 12, 725882-725882 (2021-11-06)
Monoclonal antibodies (mAbs) are promising alternatives to treat infectious diseases, especially given their potential for applications in combination therapies with antimicrobial drugs to enhance the antifungal efficacy. Protection mediated by mAbs used to treat experimental paracoccidioidomycosis (PCM) has been demonstrated
Huan Long et al.
BMC neuroscience, 12, 17-17 (2011-02-08)
Disabled-1 (Dab1) is an adaptor protein that is essential for the intracellular transduction of Reelin signaling, which regulates the migration and differentiation of postmitotic neurons during brain development in vertebrates. Dab1 function depends on its tyrosine phosphorylation by Src family
Allison L Fisher et al.
The Journal of biological chemistry, 297(4), 101156-101156 (2021-09-05)
In plasma, iron is normally bound to transferrin, the principal protein in blood responsible for binding and transporting iron throughout the body. However, in conditions of iron overload when the iron-binding capacity of transferrin is exceeded, non-transferrin-bound iron (NTBI) appears

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service