All Photos(3)
About This Item
Empirical Formula (Hill Notation):
C3H4N2 · HCl
CAS Number:
Molecular Weight:
104.54
Beilstein:
3678283
MDL number:
UNSPSC Code:
12161700
eCl@ss:
39161001
PubChem Substance ID:
NACRES:
NA.25
Recommended Products
Assay
≥98% (TLC)
≥98.5% (perchloric acid titration)
form
powder
pH
9.0-10.0
mp
158-161 °C (lit.)
solubility
methanol: 50 mg/mL, clear, colorless to faintly yellow
application(s)
diagnostic assay manufacturing
SMILES string
Cl.c1c[nH]cn1
InChI
1S/C3H4N2.ClH/c1-2-5-3-4-1;/h1-3H,(H,4,5);1H
InChI key
JDIIGWSSTNUWGK-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Imidazole hydrochloride has been used:
- in double immunohistochemistry
- to determine the activity of octopine dehydrogenase (ODH) in Mytilus galloprovincialis
- to study the cytoskeletal features during in vitro fertilization after intracytoplasmic sperm injection (ICSI) among humans
- as a reagent in the synthesis of a probe for monitoring herpes simplex virus type 1 thymidine kinase (HSV1-tk) reporter gene expression in rodents
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Exploring the cytoskeleton during intracytoplasmic sperm injection in humans
Microinjection: Methods and Applications, 189-206 (2009)
The alpha4beta2 nicotine acetylcholine receptor agonist is pronicline induces c-Fos expression in selective regions of the rat forebrain
Jacobsen J, et al.
Neuroscience Letters, 515(1), 7-11 (2012)
Biochemical responses of the marine mussel Mytilus galloprovincialis to petrochemical environmental contamination along the North-western coast of Portugal
Lima I, et al.
Chemosphere, 66(7), 1230-1242 (2007)
Synthesis of a probe for monitoring HSV1-tk reporter gene expression using chemical exchange saturation transfer MRI.
Bar-Shir A
Nature Protocols, 8(12), 2380-2380 (2013)
Jin-Woo Kim et al.
Biotechnology for biofuels, 9, 265-265 (2016-12-19)
2,3-Butanediol (2,3-BD) is a promising compound for various applications in chemical, cosmetic, and agricultural industries. Pyruvate decarboxylase (Pdc)-deficient Saccharomyces cerevisiae is an attractive host strain for producing 2,3-BD because a large amount of pyruvate could be shunted to 2,3-BD production
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service