Skip to Content
Merck
All Photos(1)

Key Documents

I1646

Sigma-Aldrich

IL-3 human

≥97% (HPLC and SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture

Synonym(s):

hIL-3, IL-3

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

product name

Interleukin-3 human, IL-3, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture

biological source

human

Quality Level

recombinant

expressed in E. coli

Assay

≥97% (HPLC and SDS-PAGE)

form

lyophilized powder

potency

≤1.0 ng/mL ED50/EC50

quality

endotoxin tested

mol wt

15 kDa

packaging

pkg of 10 and 50 μg

storage condition

avoid repeated freeze/thaw cycles

technique(s)

cell culture | mammalian: suitable

impurities

≤0.1 EU/μg

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... IL3(3562)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Interleukin-3 (IL-3) is a multifunctional protein, originally called colony forming unit-stimulating activity (CFU-SA), and is produced by activated T lymphocytes. IL-3 supports the formation of multilineage colonies in the early development of multipotent hematopoietic progenitor cells. IL-3 has been shown to induce colony formation of macrophages, neutrophils, mast cells, and megakaryocytes from agar-suspended bone marrow cells. IL-3 also interacts with IL-2 to stimulate growth of T lymphocytes and to induce IgG secretion from activated B cells. Other synonyms for IL-3 include pan-specific hemopoietin, multicolony stimulating factor, mast cell growth factor, burst promoting activity, histamine-producing cell stimulating factor, P cell stimulating factor, and WEHI-3 factor.

Physical form

Lyophilized with no additives. Sterile filtered through a 0.2 μm filter.

Analysis Note

The biological activity is measured by the dose-dependent stimulation of the proliferation of human TF-1 cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bingrui Wang et al.
Cell discovery, 8(1), 41-41 (2022-05-10)
Ribosomal protein dysfunction causes diverse human diseases, including Diamond-Blackfan anemia (DBA). Despite the universal need for ribosomes in all cell types, the mechanisms underlying ribosomopathies, which are characterized by tissue-specific defects, are still poorly understood. In the present study, we
Ding Wang et al.
iScience, 26(6), 106917-106917 (2023-06-28)
The role of SET domain containing 7 (SETD7) during human hematopoietic development remains elusive. Here, we found that deletion of SETD7 attenuated the generation of hematopoietic progenitor cells (HPCs) during the induction of hematopoietic differentiation from human embryonic stem cells
S Yousefi et al.
Allergy, 70(9), 1184-1188 (2015-06-05)
Basophils are primarily associated with immunomodulatory functions in allergic diseases and parasitic infections. Recently, it has been demonstrated that both activated human and mouse basophils can form extracellular DNA traps (BETs) containing mitochondrial DNA and granule proteins. In this report
Orit Uziel et al.
Biochemical and biophysical research communications, 450(1), 274-282 (2014-06-08)
Treatment with erythropoietin (EPO) in several cancers is associated with decreased survival due to cancer progression. Due to the major importance of telomerase in cancer biology we hypothesized that some of these effects may be mediated through EPO effect on
Hai Zhou et al.
Thrombosis and haemostasis, 113(5), 1021-1034 (2015-01-09)
Impaired megakaryocyte maturation and insufficient platelet production have been shown to participate in the pathogenesis of immune thrombocytopenia (ITP). Our previous study demonstrated that low expression of tumour necrosis factor-related apoptosis-inducing ligand (TRAIL) in megakaryocytes contributed to impaired platelet production

Articles

Interleukins, released from leukocytes, include IL-1β playing crucial roles in inflammation, immune responses, and anti-tumor immunity.

Interleukins, released from leukocytes, include IL-1β playing crucial roles in inflammation, immune responses, and anti-tumor immunity.

Interleukins, released from leukocytes, include IL-1β playing crucial roles in inflammation, immune responses, and anti-tumor immunity.

Interleukins, released from leukocytes, include IL-1β playing crucial roles in inflammation, immune responses, and anti-tumor immunity.

Protocols

Stem cell reprogramming protocols to generate human induced pluripotent stem cells (iPSCs) including viral and non-viral RNA based methods.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service