Skip to Content
Merck
All Photos(1)

Key Documents

I135

Sigma-Aldrich

Imetit dihydrobromide

≥98% (HPLC)

Synonym(s):

S-[2-(Imidazol-4-yl)ethyl]isothiourea dihydrobromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10N4S · 2HBr
CAS Number:
Molecular Weight:
332.06
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 20 mg/mL, clear

SMILES string

Br[H].Br[H].NC(=N)SCCc1c[nH]cn1

InChI

1S/C6H10N4S.2BrH/c7-6(8)11-2-1-5-3-9-4-10-5;;/h3-4H,1-2H2,(H3,7,8)(H,9,10);2*1H

InChI key

DOBOYMKCRRLTRF-UHFFFAOYSA-N

Gene Information

human ... HRH3(11255)

Biochem/physiol Actions

Potent and selective H3 histamine receptor agonist.

Features and Benefits

This compound is featured on the Histamine Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Garbarg et al.
The Journal of pharmacology and experimental therapeutics, 263(1), 304-310 (1992-10-01)
The effects of a new agonist of histamine (HA) H3 receptors, Imetit (S-[2-(4-(imidazolyl)ethyl]isothiourea) were investigated in vitro and in vivo and compared to those of (R)-alpha-methylhistamine [(R)-alpha-MeHA], a prototypic drug. Imetit inhibited the binding of [3H](R-alpha-MeHA to rat brain membranes
Balasubramaniam Annamalai et al.
ACS chemical neuroscience, 11(3), 466-476 (2020-01-10)
Reuptake and clearance of released serotonin (5-HT) are critical in serotonergic neurotransmission. Serotonin transporter (SERT) is mainly responsible for clearing the extracellular 5-HT. Controlled trafficking, phosphorylation, and protein stability have been attributed to robust SERT activity. H3 histamine receptors (H3Rs)
Carina Höring et al.
International journal of molecular sciences, 21(22) (2020-11-14)
In drug discovery, assays with proximal readout are of great importance to study target-specific effects of potential drug candidates. In the field of G protein-coupled receptors (GPCRs), the determination of GPCR-G protein interactions and G protein activation by means of
Nayeli Rivera-Ramírez et al.
Neurochemistry international, 101, 38-47 (2016-11-05)
The histamine H3 receptor (H3R) is abundantly expressed in the Central Nervous System where it regulates several functions pre and postsynaptically. H3Rs couple to Gαi/o proteins and trigger or modulate several intracellular signaling pathways, including the cAMP/PKA pathway and the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service