Skip to Content
Merck
All Photos(1)

Documents

HT80116

Sigma-Aldrich

Carbol-Fuchsin Solution

Synonym(s):

4,4′-[(4-iminocyclohexa-2,5-dien-1-ylidene)methylene]bis(2-methylaniline) hydrochloride, Pararosaniline

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116121
PubChem Substance ID:
NACRES:
NA.47

form

solution

Quality Level

IVD

for in vitro diagnostic use

pH

4.4

application(s)

hematology
histology

storage temp.

room temp

InChI

1S/C21H21N3.ClH/c1-13-11-16(5-9-19(13)23)21(15-3-7-18(22)8-4-15)17-6-10-20(24)14(2)12-17;/h3-12,22H,23-24H2,1-2H3;1H

InChI key

YCIVZWKFFBDLTG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Intended for use in the Acid Fast Stain, Procedure No. HT80 with Malachite Green counterstain Catalog No. HT80-2

Other Notes

Pararosaniline (certified), 0.85%; phenol, 2.5%; glycerol, 5%; DMSO, 5%, and ethyl alcohol, 5%, in deionized water

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Carc. 1B - Eye Irrit. 2 - Muta. 2 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

An overview of the science and practice of bacteriology in clinical diagnostics. Learn more about the application of standard and special stains for microscopic analysis.

An overview of the science and practice of bacteriology in clinical diagnostics. Learn more about the application of standard and special stains for microscopic analysis.

An overview of the science and practice of bacteriology in clinical diagnostics. Learn more about the application of standard and special stains for microscopic analysis.

An overview of the science and practice of bacteriology in clinical diagnostics. Learn more about the application of standard and special stains for microscopic analysis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service