Skip to Content
Merck
All Photos(2)

Key Documents

HT153

Sigma-Aldrich

Phosphomolybdic Acid Solution

Synonym(s):

Phosphomolybdic acid solution, Molybdophosphoric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H3[P(Mo3O10)4]
CAS Number:
Molecular Weight:
1825.25
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.47
Pricing and availability is not currently available.

form

solution

Quality Level

shelf life

Expiry date on the label

reaction suitability

reagent type: oxidant

IVD

for in vitro diagnostic use

concentration

10%

application(s)

hematology
histology

storage temp.

room temp

SMILES string

O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.OP(O)(O)=O

InChI

1S/12Mo.H3O4P.36O/c;;;;;;;;;;;;1-5(2,3)4;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;/h;;;;;;;;;;;;(H3,1,2,3,4);;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;

InChI key

DHRLEVQXOMLTIM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Intended for use in the Sigma Masson Trichrome Procedure, HT15.

Components

Phosphomolydic Acid, 10%. Formation of precipitate does not affect performance in the Masson Trichrome Procedure.

Pictograms

Flame over circleCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Ox. Liq. 3 - Skin Corr. 1B

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrey V Bulatov et al.
Talanta, 96, 62-67 (2012-07-24)
Simple, rapid and fully automated methods for the manual and automated spectrophotometric determination of epinephrine have been developed by using schemes of stepwise injection (SWIA) and sequential injection analysis (SIA) implemented in the same manifold. The determination is based on
Juan Feng et al.
Analytical biochemistry, 409(1), 144-149 (2010-10-26)
The classical malachite green (MLG) assay of phosphate, which added MLG after molybdate to the acidified reaction solutions of phosphate, tolerated interference from papaverine, sildenafil, and some similar hydrophobic amines. Resonance Rayleigh scattering signals, the alleviation of interference by poly(vinyl
Paweł K Zarzycki et al.
Analytica chimica acta, 688(2), 168-174 (2011-02-22)
The main goal of present paper is to demonstrate the separation and detection capability of micro-TLC technique involving simple one step liquid extraction protocols of complex materials without multi-steps sample pre-purification. In the present studies target components (cyanobacteria pigments, lipids
Maher Mahmoud El-Hashash et al.
Acta pharmaceutica (Zagreb, Croatia), 60(3), 361-367 (2010-12-08)
In the present study, antioxidant activity of methanolic extracts of the leaves of seven Egyptian Cassia species was investigated using two methods, the phosphomolyb-date method and 1,1 diphenyl-2-picrylhydrazyl radical (DPPH) scavenging activity method.· The results revealed that C. glauca is
Dawei Fan et al.
Journal of colloid and interface science, 351(1), 151-155 (2010-08-19)
Hybrid films composed of oppositely charged Keggin-type 12-molybdophosphoric acid H(3)PMo(12)O(40).nH(2)O (PMo(12)) and water-soluble cationic meso-tetra (N-methyl-4-pyridyl) porphinetetratosylate (TMPyP) were fabricated onto silicon and ITO substrates by a layer-by-layer (LbL) self-assembly technique. The LbL films were characterized by UV-vis spectroscopy, X-ray

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service