Skip to Content
Merck
All Photos(5)

Key Documents

HPA047819

Sigma-Aldrich

Anti-PPP1R17 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-C7orf16, Anti-GSBS, Anti-protein phosphatase 1, regulatory subunit 17

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:1000-1:2500

immunogen sequence

DRLDKLDPRCSHLDDLSDQFIKDCDLKKKPRKGKNVQATLNVESDQKKPRRKDTPALH

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PPP1R17(10842)

General description

The gene PPP1R17 (protein phosphatase 1 regulatory subunit 17) is mapped to human chromosome 7p15. The gene encodes for a cGMP (cyclic guanosine monophosphate) dependent protein kinase. It is present in Purkinje cells of the cerebellum.

Immunogen

protein phosphatase 1, regulatory subunit 17 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies®Powered by Atlas Antibodies is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-PPP1R17 antibody produced in rabbit has been used in immunohistochemistry.

Biochem/physiol Actions

PPP1R17 (protein phosphatase 1 regulatory subunit 17) might be associated with long-term depression. Presence of PPP1R17 in the hypothalamus might also be associated with food intake via the hypothalamo-pituitary-adrenal axis. PPP1R17 variants are linked with hypercholesterolemia.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST78462

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A promoter SNP (-1323T>C) in G-substrate gene (GSBS) correlates with hypercholesterolemia.
Ono S, et al.
Journal of Human Genetics, 48, 447-450 (2003)
Linkage of genes to total lean body mass in normal women.
Livshits G, et al.
The Journal of Clinical Endocrinology and Metabolism, 92, 3171-3176 (2007)
Comprehensive Classification of Retinal Bipolar Neurons by Single-Cell Transcriptomics.
Shekhar K, et al.
Cell, 166, 1308-1323 (2016)
William N Grimes et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2021-06-05)
Amacrine cells are interneurons composing the most diverse cell class in the mammalian retina. They help encode visual features such as edges or directed motion by mediating excitatory and inhibitory interactions between input (i.e. bipolar) and output (i.e. ganglion) neurons
Andrew Jo et al.
Nature communications, 14(1), 7746-7746 (2023-11-27)
Neural computations arise from highly precise connections between specific types of neurons. Retinal ganglion cells (RGCs) with similar stratification patterns are positioned to receive similar inputs but often display different response properties. In this study, we used intersectional mouse genetics

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service