Skip to Content
Merck
All Photos(3)

Key Documents

HPA044144

Sigma-Aldrich

Anti-WDPCP antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-BBS15, Anti-C2orf86, Anti-Fritz, Anti-WD Repeat Containing Planar Cell Polarity Effector, Anti-hFrtz

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
conjugate:
unconjugated
application:
IF
IHC
clone:
polyclonal
species reactivity:
human
citations:
5
technique(s):
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

DIIFQYIHCDEIYEAINILSSMNWDTLGHQCFISMSAIVNHLLRQKLTPEREAQLETSLGTFYAPTRPLLDSTILEYRDQISKYARRF

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... WDPCP(51057)

General description

WD repeat containing planar cell polarity effector (WDPCP) was previously known as FRITZ. WDPCP is located on human chromosome location 2p15. WDPCP is located in the cytoplasm of focal adhesions and actin cytoskeletons.

Immunogen

WD repeat containing planar cell polarity effector recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

WDPCP has major importance in maintaining the stability of basolateral plasma membrane of epithelial tissues. It also regulates the cortical tension and rigidity of epithelial cells. It is an important protein necessary for ciliogenesis and acts as an effector for planar cell polarity signalling system. Inflammatory cytokines such as type1 T helper cells (Th1) reduces the expression of WDPCP which causes poor quantity and length of cilia and also reduces Septin7.WDPCP also helps in collective cell movement. WDPCP promotes remodelling of coronary primitive plexus by regulating Epithelial to mesenchymal transition (EMT) and epicardium-derived cells (EPDC) migration. Mutations in this protein causes Bardet-Biedl/Meckel-Gruber ciliopathy syndromes.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST83410

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The planar cell polarity effector protein Wdpcp (Fritz) controls epithelial cell cortex dynamics via septins and actomyosin
Park, Tae, et al.
Biochemical and Biophysical Research Communications, 456(2), 562-566 (2015)
Pathogenic variants screening in seventeen candidate genes on 2p15 for association with ankylosing spondylitis in a Han Chinese population
Wang, Men, et al.
Testing, 12(5), e0177080-e0177080 (2017)
WDPCP regulates the ciliogenesis of human sinonasal epithelial cells in chronic rhinosinusitis
Ma, Yun a, et al.
Cytoskeleton (Hoboken, N.J.), 74(2), 82-90 (2017)
Wdpcp promotes epicardial EMT and epicardium-derived cell migration to facilitate coronary artery remodeling
Liu, Xian, et al.
Science Signaling, 11(519), eaah5770-eaah5770 (2018)
Cheng Cui et al.
PLoS biology, 11(11), e1001720-e1001720 (2013-12-05)
Planar cell polarity (PCP) regulates cell alignment required for collective cell movement during embryonic development. This requires PCP/PCP effector proteins, some of which also play essential roles in ciliogenesis, highlighting the long-standing question of the role of the cilium in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service