Skip to Content
Merck
All Photos(5)

Key Documents

HPA036774

Sigma-Aldrich

Anti-NKX6-1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-NK6 homeobox 1, Anti-NKX6A, Anti-Nkx6.1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

PLGTHNPGGLKPPATGGLSSLGSPPQQLSAATPHGINDILSRPSM

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NKX6-1(4825)

Related Categories

General description

NKX6-1 (NK6 homeobox 1), a homeobox transcription factor is coded by NKX6A gene. It is located on human chromosome 4q21.2-q22. NKX6-1 belongs to the NK homeobox gene family. It is specific in adult cells producing insulin.

Immunogen

NK6 homeobox 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry. The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-NKX6-1 antibody has been used in immunofluorescence, FAIRE (formaldehyde-assisted isolation of regulatory elements) and ChIP and immunohistochemistry.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Chromatin immunoprecipitation (1 paper)

Biochem/physiol Actions

NKX6-1 (NK6 homeobox 1) is involved in the growth and modulation of endocrine activities of pancreatic islets. It acts as an immunohistochemical marker for pancreatic and duodenal neuroendocrine tumors. NKX6-1 also participates in lineage specification in early pancreatic progenitor tissue. This gene regulates the expression of several pancreas-related genes.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST80893

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Noémie Mlp Bérenger-Currias et al.
Journal of tissue engineering, 13, 20417314221103042-20417314221103042 (2022-06-17)
Stem-cell derived in vitro systems, such as organoids or embryoids, hold great potential for modeling in vivo development. Full control over their initial composition, scalability, and easily measurable dynamics make those systems useful for studying specific developmental processes in isolation.
Duo An et al.
Proceedings of the National Academy of Sciences of the United States of America, 115(2), E263-E272 (2017-12-28)
Cell encapsulation has been shown to hold promise for effective, long-term treatment of type 1 diabetes (T1D). However, challenges remain for its clinical applications. For example, there is an unmet need for an encapsulation system that is capable of delivering
Isolation, characterization, and chromosomal mapping of the human Nkx6. 1 gene (NKX6A), a new pancreatic islet homeobox gene.
Inoue H, et al.
Genomics, 40(2), 367-370 (1997)
Generation of thalamic neurons from mouse embryonic stem cells.
Shiraishi A, et al.
Development, 144, 1211-1220 (2017)
Minghui Wu et al.
Cell reports, 37(5), 109919-109919 (2021-11-04)
Type 2 diabetes mellitus (T2D) is a chronic age-related disorder characterized by hyperglycemia due to the failure of pancreatic beta cells to compensate for increased insulin demand. Despite decades of research, the pathogenic mechanisms underlying T2D remain poorly defined. Here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service