Skip to Content
Merck
All Photos(3)

Key Documents

HPA024604

Sigma-Aldrich

Anti-FAM83H antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2

Synonym(s):

Anti-Protein FAM83H

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

VVSQAWREEVAAPGAVGGERRSLESCLLDLRDSFAQQLHQEAERQPGAASLTAAQLLDTLGRSGSDRLPSRFLSAQSHSTSPQGLDSPLPLEGS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FAM83H(286077)

General description

The gene FAM83H (family with sequence similarity 83 member H) is mapped to human chromosome 8q24.3. The protein localizes with the trans Golgi network.

Immunogen

Protein FAM83H recombinant protein epitope signature tag (PrEST)

Application

Anti-FAM83H antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

FAM83H (family with sequence similarity 83 member H) is upregulated in various cancers. In colorectal cancer, it regulates the dynamics of keratin cytoskeleton. Mutations in it are associated with hypocalcified amelogenesis imperfecta.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST76417

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dorte Haubek et al.
International journal of paediatric dentistry, 21(6), 407-412 (2011-06-28)
BACKGROUND.  Autosomal dominant hypocalcified amelogenesis imperfecta (ADHCAI) is a disease with severe dental manifestations. OBJECTIVES.  The aims were by means of a genome-wide linkage scan to search for the gene underlying the ADHCAI phenotype in a Danish five-generation family and
Teruki Yanagi et al.
Cancer research, 78(24), 6795-6806 (2018-11-06)
: TRIM29 (tripartite motif-containing protein 29) is a TRIM family protein that has been implicated in breast, colorectal, and pancreatic cancers. However, its role in stratified squamous epithelial cells and tumors has not been elucidated. Here, we investigate the expression
Takahisa Kuga et al.
Journal of cell science, 126(Pt 20), 4721-4731 (2013-08-02)
Keratin filaments form cytoskeletal networks in epithelial cells. Dynamic rearrangement of keratin filament networks is required for epithelial cells to perform cellular processes such as cell migration and polarization; however, the mechanism governing keratin filament rearrangement remains unclear. Here, we
Takahisa Kuga et al.
Scientific reports, 12(1), 11819-11819 (2022-07-14)
The casein kinase 1 (CK1) family of serine/threonine protein kinases is involved in diverse cellular events at discrete subcellular compartments. FAM83H acts as a scaffold protein that recruits CK1 to the keratin cytoskeleton or to the nuclear speckles, which are
Takahisa Kuga et al.
Scientific reports, 6, 34472-34472 (2016-09-30)
In some fibroblasts, casein kinase 1α (CK1α) is localized to nuclear speckles, which are sub-nuclear compartments supplying splicing factors, whereas it is recruited on keratin filaments in colorectal cancer cells such as DLD1 cells. In order to obtain a deeper

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service