Skip to Content
Merck
All Photos(1)

Key Documents

HPA019556

Sigma-Aldrich

Anti-ABCG8 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-ATP-binding cassette sub-family G member 8, Anti-Sterolin-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

HMVQYFTAIGYPCPRYSNPADFYVDLTSIDRRSREQELATREKAQSLAALFLEKVRDLDDFLWKAETKDLDEDTCVESSVTPLDTNCLPSPTKMPGAVQQFTTLIRRQISNDFRDLP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ABCG8(64241)

General description

ABCG8 (ATP-binding cassette, sub-family G (WHITE), member 8) is an ATP-binding cassette (ABC) transporter belonging to the ABC transporter family. It is expressed in the liver and small intestine.

Immunogen

ATP-binding cassette sub-family G member 8 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

ABCG8 (ATP-binding cassette, sub-family G (WHITE), member 8) is involved in the cholesterol homeostasis via several regulatory pathways. It maintains sterol level by sterol absorption and by promoting biliary sterol excretion. In conjugation with another ABC transporter (ABCG5), ABCG8 acts as a heterodimer. It also performs a vital role in the enterohepatic trafficking of cholesterol, which further alters the rate of biliary sterol excretion. Mutation in ABCG8 causes an autosomal recessive disorder of sterol metabolism, sitosterolemia.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73218

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joyce J Repa et al.
The Journal of biological chemistry, 277(21), 18793-18800 (2002-03-20)
Mutations in the ATP-binding cassette (ABC) transporters ABCG5 and ABCG8 have recently been shown to cause the autosomal recessive disorder sitosterolemia. Here we demonstrate that the ABCG5 and ABCG8 genes are direct targets of the oxysterol receptors liver X receptor
Sang Bong Ahn et al.
The Korean journal of internal medicine, 34(4), 777-784 (2018-02-23)
Intestinal cholesterol absorption includes intestinal Niemann-Pick C1-like 1 (NPC1L1) and is an important target pathway in nonalcoholic fatty liver disease (NAFLD). We investigated the expression of NPC1L1 and its correlation with liver X receptor (LXR) expression in peripheral mononuclear (PMN)
Zixun Yang et al.
Journal of agricultural and food chemistry, 67(20), 5782-5791 (2019-05-06)
Reverse cholesterol transport (RCT) is a physiological process, in which excess peripheral cholesterol is transported to the liver and further excreted into the bile and then feces. Recently, fucoidans are reported to have a lipid-lowering effect. This study was designed
Liqing Yu et al.
The Journal of clinical investigation, 110(5), 671-680 (2002-09-05)
Two ATP-binding cassette (ABC) transporters, ABCG5 and ABCG8, have been proposed to limit sterol absorption and to promote biliary sterol excretion in humans. To test this hypothesis, a P1 clone containing the human ABCG5 and ABCG8 genes was used to
Ruoqi Li et al.
Biochimica et biophysica acta. Molecular and cell biology of lipids, 1864(8), 1124-1133 (2019-05-06)
Hypercholesterolemia is a preventable risk factor for atherosclerosis and cardiovascular disease. However, the mechanisms of diosgenin (DG) that promote cholesterol homeostasis and alleviate hypercholesterolemia remain elusive. To investigate the effects and molecular mechanisms of the promotion of cholesterol metabolism by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service