Skip to Content
Merck
All Photos(1)

Key Documents

HPA019511

Sigma-Aldrich

Anti-SLIT2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1

Synonym(s):

Anti-Slit homolog 2 protein, Anti-Slit-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

SAIYSVETINDGNFHIVELLALDQSLSLSVDGGNPKIITNLSKQSTLNFDSPLYVGGMPGKSNVASLRQAPGQNGTSFHGRIWN

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLIT2(9353)

General description

SLIT2 (slit homolog 2) is an extracellular axon guidance molecule belonging to the Slit/Roundabout (Robo) family. It is expressed in human placenta.

Immunogen

Slit homolog 2 protein Precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

SLIT2 (slit homolog 2) is majorly involved in the regulation of inflammatory response. It plays a key role in pro-inflammatory cytokines in placental nutrient transport. It acts as the neuronal guidance protein to suppress the fibrocyte differentiation. It directly binds to the Robo receptors as repulsive ligands. It has been reported that it can be used as a therapeutic for fibrosis by controlling Slit2 signaling.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74496

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ratana Lim et al.
American journal of reproductive immunology (New York, N.Y. : 1989), 73(1), 66-78 (2014-10-21)
Obese pregnancies are characterised by increased inflammation. Members of the Slit/Roundabout (Robo) family are key regulators of the inflammatory response. The aim of this study was to determine the effect of (i) pre-existing maternal obesity on Slit-Robo expression in human
Qin Chen et al.
Experimental and therapeutic medicine, 22(3), 1042-1042 (2021-08-11)
Sepsis and septic shock are the main cause of mortality in intensive care units. The prevention and treatment of sepsis remains a significant challenge worldwide. The endothelial cell barrier plays a critical role in the development of sepsis. Aminophylline, a
K Brose et al.
Cell, 96(6), 795-806 (1999-04-02)
Extending axons in the developing nervous system are guided in part by repulsive cues. Genetic analysis in Drosophila, reported in a companion to this paper, identifies the Slit protein as a candidate ligand for the repulsive guidance receptor Roundabout (Robo).
Darrell Pilling et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(51), 18291-18296 (2014-12-10)
Monocytes leave the blood and enter tissues. In healing wounds and fibrotic lesions, some of the monocytes differentiate into fibroblast-like cells called fibrocytes. In healthy tissues, even though monocytes enter the tissue, for unknown reasons, very few monocytes differentiate into

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service