Skip to Content
Merck
All Photos(2)

Key Documents

HPA019070

Sigma-Aldrich

Anti-AHNAK antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Desmoyokin, Anti-Neuroblast differentiation-associated protein AHNAK

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

DIKSPSLDVTVPEAELNLETPEISVGGKGKKSKFKMPKIHMSGPKIKAKKQGFDLNVPGGEIDASLKAPDVDVNIAGPDAALKVDVKSPKTKKTMFGKMYFPDVEFDIKSPKFKAEAPLPSPKL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AHNAK(79026)

General description

The gene AHNAK (neuroblast differentiation-associated protein) is mapped to human chromosome 11q12.2. AHNAK is strongly expressed in muscular cells, for instance cardiomyocytes and skeletal muscle cells. The protein localizes at the plasma membrane, cytoplasm and nucleus. AHNAK is commonly called as desmoyokin.

Immunogen

Neuroblast differentiation-associated protein AHNAK recombinant protein epitope signature tag (PrEST)

Application

Anti-AHNAK antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

AHNAK (neuroblast differentiation-associated protein) interacts with muscle membrane repair protein, Dysferlin. It is recruited to the bacterial invasion site and is involved in type-3 secretion system mediated invasion of Salmonella and Chlamydia trachomatis. AHNAK is down-regulated in neuroblastoma cell line, small cell lung carcinomas and Burkitt lymphomas, whereas, up-regulated in prostate cancer cell line, breast cancer, fibrosarcoma and glioma.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74734

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Claudia A Dumitru et al.
PloS one, 8(2), e56420-e56420 (2013-02-15)
AHNAK/Desmoyokin is a giant protein which has been recently linked to reorganization of the actin cytoskeleton, cellular migration and invasion. Here, we investigated the role of AHNAK in the pathophysiology of larynx carcinoma-one of the major subtypes of head and
Hitomi Sudo et al.
International journal of oncology, 44(2), 530-538 (2013-11-21)
The worldwide incidence of the highly aggressive tumor mesothelioma is expected to increase. Mesothelioma is classified into three main histological subtypes: epithelioid, sarcomatoid and biphasic. Although the pathological diagnostic markers for epithelioid are established, to date no adequate marker for
P D Kingsley et al.
Development, growth & differentiation, 43(2), 133-143 (2001-04-04)
The gene product ahnak has been identified from extra-embryonic mesoderm cDNA enriched using a subtractive hybridization approach modified for using small amounts of starting material. Clones for cyclin D2 and H19 have also been isolated as being preferentially enriched in
Carrie Jolly et al.
Cellular microbiology, 16(1), 64-77 (2013-08-13)
The facultative intracellular pathogen, Salmonella enterica, triggers its own uptake into non-phagocytic epithelial cells. Invasion is dependent on a type 3 secretion system (T3SS), which delivers a cohort of effector proteins across the plasma membrane where they induce dynamic actin-driven
Yanchao Huang et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 21(3), 732-742 (2006-12-23)
Mutations in dysferlin cause limb girdle muscular dystrophy 2B, Miyoshi myopathy and distal anterior compartment myopathy. Dysferlin is proposed to play a role in muscle membrane repair. To gain functional insight into the molecular mechanisms of dysferlin, we have searched

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service