Skip to Content
Merck
All Photos(14)

Key Documents

HPA019025

Sigma-Aldrich

Anti-SERPINB5 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Maspin, Anti-Protease inhibitor 5, Anti-Serpin B5

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

rat, human

enhanced validation

RNAi knockdown
independent
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

MDALQLANSAFAVDLFKQLCEKEPLGNVLFSPICLSTSLSLAQVGAKGDTANEIGQVLHFENVKDVPFGFQTVTSDVNKLSSFYSLKLIKRLYVD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SERPINB5(5268)

General description

The gene SERPINB5 (serpin peptidase inhibitor, clade B, member 5) is mapped to human chromosome 18q21.33. The protein localizes in the cytoplasm and the nucleus. SERPINB5 is generally referred as MASPIN (mammary serine protease inhibitor).

Immunogen

Serpin B5 Precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

SERPINB5 (serpin peptidase inhibitor, clade B, member 5) is a class II tumor suppressor gene. It can mediate apoptosis and suppress cell invasion. It also inhibits neovascularization and decreases tumor-linked microvessels. It is down-regulated in breast cancer cells, esophageal squamous cell carcinoma, glioma cells and lung adenocarcinoma.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74611

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fulvio Lonardo et al.
Histopathology, 65(6), 757-763 (2014-07-22)
To test whether changes in the subcellular localization of maspin parallel morphological progression in pulmonary adenocarcinoma, we compared its expression between lepidic and invasive growth patterns. Applying immunohistochemistry, we compared maspin expression in lepidic and invasive growth patterns occurring in
Siqi Ma et al.
Pakistan journal of pharmaceutical sciences, 28(3 Suppl), 1059-1063 (2015-06-09)
This paper tested and analyzed the expression of ATF3 (activating transcription factor), MMP-2 (matrix metalloprotease) and maspin in tissue chip of glioma and its correlation with glioma advancement. Based on immunohistochemical staining, this paper selected 100 patients with glioma and
Hui Meng et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 36(8), 6231-6238 (2015-03-18)
Esophageal squamous cell carcinoma (ESCC) is one of the most aggressive cancers in the world. Epidemiological survey studies have verified that the development of ESCC relates to a complex interactive process between multiple genetic susceptibilities and environmental exposure. Serpins are
Da Eun Lee et al.
PloS one, 8(11), e78136-e78136 (2013-11-14)
Quantification of cell-free fetal DNA by methylation-based DNA discrimination has been used in non-invasive prenatal testing of fetal chromosomal aneuploidy. The maspin (Serpin peptidase inhibitor, clade B (ovalbumin), member 5; SERPINB5) gene, located on chromosome 18q21.33, is hypomethylated in the
Caleb M Bailey et al.
The Journal of biological chemistry, 280(40), 34210-34217 (2005-07-29)
Since its reported discovery in 1994, maspin (mammary serine protease inhibitor) has been characterized as a class II tumor suppressor by its ability to promote apoptosis and inhibit cell invasion. Maspin is highly expressed in normal mammary epithelial cells but

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service