Skip to Content
Merck
All Photos(3)

Key Documents

HPA019002

Sigma-Aldrich

Anti-IPO7 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Imp7, Anti-Importin-7, Anti-Ran-binding protein 7, Anti-RanBP7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

mouse, rat, human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

RAYACHAEHENDSDDDDEAEDDDETEELGSDEDDIDEDGQEYLEILAKQAGEDGDDEDWEEDDAEETALEGYSTIIDDEDNPVDEY

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IPO7(10527)

General description

Importin-7 (IPO7) localizes in the cytoplasm and the nucleus.

Immunogen

Importin-7 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Importin-7 (IPO7) is involved in nuclear import of proteins in Ran-dependent manner. It can form heterodimer with importin β. Importin-7 is crucial for import of ribosomal proteins, histone H1, HIV-1 (Human immunodeficiency virus 1) intracellular reverse transcription complexes, early growth response-1, hypoxia-inducible factor (HIF-1α), component of human telomerase, glioma-associated oncogene homolog-1 (GLI1), supercoiled plasmid DNA and human mitochondrial DNA.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73988

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cornelia Frohnert et al.
PloS one, 9(2), e88887-e88887 (2014-03-04)
In actively dividing eukaryotic cells, chromosome ends (telomeres) are subject to progressive shortening, unless they are maintained by the action of telomerase, a dedicated enzyme that adds DNA sequence repeats to chromosomal 3'end. For its enzymatic function on telomeres, telomerase
Georgia Chachami et al.
Biochemical and biophysical research communications, 390(2), 235-240 (2009-10-01)
Hypoxia-inducible transcription factor 1 (HIF-1) mediates the cellular response to hypoxia. HIF-1 activity is controlled via the synthesis, degradation or intracellular localization of its alpha subunit. HIF-1alpha contains a C-terminal bipartite basic NLS that interacts with importins alpha. We have
Li Ni et al.
Molecular and cellular biology, 33(24), 4766-4778 (2013-10-09)
The androgen receptor (AR) has critical functions as a transcription factor in both normal and cancer cells, but the specific mechanisms that regulate its nuclear localization are not well defined. We found that an AR mutation commonly reported in prostate
S Jäkel et al.
The EMBO journal, 17(15), 4491-4502 (1998-08-04)
The assembly of eukaryotic ribosomal subunits takes place in the nucleolus and requires nuclear import of ribosomal proteins. We have studied this import in a mammalian system and found that the classical nuclear import pathway using the importin alpha/beta heterodimer
S Jäkel et al.
The EMBO journal, 18(9), 2411-2423 (1999-05-06)
Import of proteins into the nucleus proceeds through nuclear pore complexes and is largely mediated by nuclear transport receptors of the importin beta family that use direct RanGTP-binding to regulate the interaction with their cargoes. We investigated nuclear import of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service