Skip to Content
Merck
All Photos(3)

Key Documents

HPA018816

Sigma-Aldrich

Anti-ANGPT1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2

Synonym(s):

Anti-ANG-1, Anti-Angiopoietin-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

SNQRRSPENSGRRYNRIQHGQCAYTFILPEHDGNCRESTTDQYNTNALQRDAPHVEPDFSSQKLQHLEHVME

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ANGPT1(284)

General description

The gene angiopoietin-1 (ANGPT1) is mapped to human chromosome 8q23.1. It belongs to angiopoietin family of growth factors. The protein has coiled-coil domains in the amino-terminus and a fibrinogen-like domain in the carboxyl-terminus.

Immunogen

Angiopoietin-1 Precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-ANGPT1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

Angiopoietin-1 (ANGPT1) is an angiogenic factor. It is crucial for normal vascular development. ANGPT1 strengthens endothelial cell junctions and protects endothelial cells from AGE (advanced glycation end product)-induced death. Serum ANGPT1 levels are lower in asthmatic children, which was proposed to be a cause of destabilization of blood vessels in the patients.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74500

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Burcu Tahire Köksal et al.
Allergy and asthma proceedings, 35(6), 482-488 (2015-01-15)
Asthma is characterized by chronic airway inflammation that is associated with structural changes termed airway remodeling. Recently, cytokines/mediators that augment inflammation have been attracting attention in this field. The aim of this study was to evaluate serum angiopoietin (Ang)-1, Ang-2
P C Maisonpierre et al.
Science (New York, N.Y.), 277(5322), 55-60 (1997-07-04)
Angiogenesis is thought to depend on a precise balance of positive and negative regulation. Angiopoietin-1 (Ang1) is an angiogenic factor that signals through the endothelial cell-specific Tie2 receptor tyrosine kinase. Like vascular endothelial growth factor, Ang1 is essential for normal
Jingling Zhao et al.
Journal of cellular physiology, 230(8), 1895-1905 (2014-12-30)
Endothelial dysfunction is a major characteristic of diabetic vasculopathy. Protection of the vascular endothelium is an essential aspect of preventing and treating diabetic vascular complications. Although Angiopoietin-1 (Ang-1) is an important endothelial-specific protective factor, whether Ang-1 protects vascular cells undergoing
John R Zibert et al.
The Journal of clinical investigation, 121(1), 410-421 (2010-12-08)
Dysregulated angiogenesis is a hallmark of chronic inflammatory diseases, including psoriasis, a common skin disorder that affects approximately 2% of the population. Studying both human psoriasis in 2 complementary xenotransplantation models and psoriasis-like skin lesions in transgenic mice with epidermal
Rui Zhang et al.
Oncology letters, 8(3), 1001-1008 (2014-08-15)
Acute promyelocytic leukemia (APL) is characterized by the t(15;17)(q22;q21), which results in the fusion of the promyelocytic leukemia (

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service