Skip to Content
Merck
All Photos(5)

Key Documents

HPA018471

Sigma-Aldrich

Anti-PDCD5 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Programmed cell death protein 5, Anti-Protein TFAR19, Anti-TF-1 cell apoptosis-related protein 19

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

MADEELEALRRQRLAELQAKHGDPGDAAQQEAKHREAEMRNSILAQVLDQSARARLSNLALV

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PDCD5(9141)

General description

The gene programmed cell death-5 (PDCD5) is mapped to human chromosome 19q13.11. The protein is mainly localized in the plasma membrane and the cytoplasm. However, under certain biological conditions PDCD5 relocalizes to the nucleus.

Immunogen

Programmed cell death protein 5 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

YY1-associated factor 2 (YAF2) binds and stabilizes programmed cell death-5 (PDCD5), thereby promoting TP53 (Tumor suppressor p53)-mediated apoptosis.[1] In lung A549 cancer cells, DNAJB1 (DnaJ homolog subfamily B member 1) binds and degrades PDCD5 to suppress p53-dependent apoptosis of cancer cells. PDCD5 stimulates promoter activity of KLF9 (Krueppel-like factor 9). Up-regulation of KLF9 inhibits cell proliferation and induces apoptosis in hepatocellular carcinoma cells. PDCD5 is down-regulated in ovarian cancer cells. Absence of PDCD5 is correlated with disease progression and stage in ovarian cancer.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74166

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li Gao et al.
Chinese medical journal, 128(8), 1084-1090 (2015-04-18)
Ovarian cancer is a leading gynecological malignancy. We investigated the prognostic value of programmed cell death 5 (PDCD5) in patients with ovarian cancer. Expression levels of PDCD5 mRNA and protein were examined in six ovarian cancer cell lines (SKOV3, CAOV3
Da-Zhi Fu et al.
International journal of oncology, 44(1), 153-160 (2013-11-01)
Liver cancer in men is the fifth most frequently diagnosed cancer worldwide. Human Krüppel-like factor (KLF9) gene, localized on human chromosome 9q13, has been implicated in mediating a diverse range of biological processes including stem cell maintenance and differentiation of
Daniel R Carrasco et al.
Cancer cell, 9(4), 313-325 (2006-04-18)
To identify genetic events underlying the genesis and progression of multiple myeloma (MM), we conducted a high-resolution analysis of recurrent copy number alterations (CNAs) and expression profiles in a collection of MM cell lines and outcome-annotated clinical specimens. Attesting to
Xiandan Cui et al.
Cancer letters, 357(1), 307-315 (2014-12-03)
Although PDCD5 promotes p53-mediated apoptosis in various cancers, little is known about PDCD5 regulation. We recently found that DNAJB1 interacts with PDCD5 and induces the ubiquitin-dependent proteasomal degradation of PDCD5, thereby inhibiting p53-mediated apoptosis. To investigate these novel roles for
Soo-Yeon Park et al.
Biochimica et biophysica acta, 1853(5), 1060-1072 (2015-01-21)
Programmed cell death 5 (PDCD5) plays a crucial role in TP53-mediated apoptosis, but the regulatory mechanism of PDCD5 itself during apoptosis remains obscure. We identified YY1-associated factor 2 (YAF2) as a novel PDCD5-interacting protein in a yeast two-hybrid screen for

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service