Skip to Content
Merck
All Photos(5)

Key Documents

HPA018015

Sigma-Aldrich

Anti-IFNAR1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-IFN-alpha-REC, Anti-Interferon-alpha/beta receptor alpha chain precursor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

KWKQIPDCENVKTTQCVFPQNVFQKGIYLLRVQASDGNNTSFWSEEIKFDTEIQAFLLPPVFNIRSLSDSFHIYIGAPKQSGNTPVIQDYPLIYEIIFWENTSNAERKIIEKKTDVTVPNLKPLTVYCVKARAHTMDEKLN

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IFNAR1(3454)

General description

Interferon-α/β receptor α chain precursor (IFNAR1) is part of a heterodimeric receptor and contains an intracellular motif with a unique peptide sequence. The gene encoding it is localized on chromosome 21.

Immunogen

Interferon-alpha/beta receptor alpha chain precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-IFNAR1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Interferon-α/β receptor α chain precursor (IFNAR1) acts as a receptor for the pro-inflammatory cytokine interferon α (IFN-α) and is involved in immune processes during malaria infection. When IFN-α binds to IFNAR1, Janus kinases (JAKs) are activated. They then phosphorylate signal transducer and activator of transcription 1 (STAT1) and STAT2. This complex then enters the nucleus and activates antiviral gene transcription. Single-nucleotide polymorphisms in IFNAR1 have shown to be risk factors for malaria. IFNAR1 is down regulated in Hepatitis C Virus (HCV)-linked chronic liver diseases.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72895

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kanika Kanchan et al.
Infection, genetics and evolution : journal of molecular epidemiology and evolutionary genetics in infectious diseases, 29, 6-14 (2014-12-03)
Pro-inflammatory cytokines IFNγ and IFNα function through their cellular receptors IFNγR1 and IFNαR1, respectively to mediate immune processes during malaria infection. A total of 21 SNPs, 2 ins/del polymorphisms and a microsatellite repeat, selected on the basis of their reported
Heidi J A Wallweber et al.
Nature structural & molecular biology, 21(5), 443-448 (2014-04-08)
Tyrosine kinase 2 (TYK2) is a member of the Janus kinase (JAK) family of nonreceptor tyrosine kinases, which are essential for proper signaling in immune responses and development. Here we present a 2.0-Å-resolution crystal structure of a receptor-binding fragment of
Olena Odnokoz et al.
Cancer biology & therapy, 21(7), 629-636 (2020-05-08)
Within the microenvironment of solid tumors, stress associated with deficit of nutrients and oxygen as well as tumor-derived factors triggers the phosphorylation-dependent degradation of the IFNAR1 chain of type I interferon (IFN1) receptor and ensuing suppression of the IFN1 pathway.
Peter M George et al.
Circulation research, 114(4), 677-688 (2013-12-18)
Evidence is increasing of a link between interferon (IFN) and pulmonary arterial hypertension (PAH). Conditions with chronically elevated endogenous IFNs such as systemic sclerosis are strongly associated with PAH. Furthermore, therapeutic use of type I IFN is associated with PAH.
R Raz et al.
Somatic cell and molecular genetics, 21(2), 139-145 (1995-03-01)
The species specificity of interferons (IFNs) depends on restricted recognition of these ligands by multisubunit cell surface receptors. Expression of the human receptor subunit IFNAR in mouse cells conferred sensitivity only to one subtype of human IFN, IFN-alpha B. Other

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service