Skip to Content
Merck
All Photos(3)

Key Documents

HPA015624

Sigma-Aldrich

Anti-GZMM antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-LMET1, Anti-MET1, Anti-granzyme M (lymphocyte met-ase 1)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

PSMVCLAADSKDQAPCKGDSGGPLVCGKGRVLAGVLSFSSRVCTDIFKPPVATAVAPYVSWIRKVTGR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GZMM(3004)

Immunogen

granzyme M (lymphocyte met-ase 1) recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Granzyme M is a unique serine protease protein encoded by the GZMM gene in humans and is expressed in cytotoxic granules of NK cells and T lymphocytes. It consists of cationic sites (cs1 and cs2), which are involved in binding to the cell surface (thereby promoting its uptake and release into the cytoplasm). It is useful in providing the first line of defense against viral pathogens and acts as an inducer of cell death. It plays an essential function for granule-mediated cytolysis and can also be helpful in inducing apoptosis of tumor cells through rapid caspase-dependent and independent pathways. GZMM plays crucial role in both innate and adaptive immunity.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72420

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lianfeng Wu et al.
Journal of immunology (Baltimore, Md. : 1950), 183(1), 421-429 (2009-06-23)
Granzyme M (GzmM), a unique serine protease constitutively expressed in NK cells, is important for granule-mediated cytolysis and can induce rapid caspase-dependent apoptosis of tumor cells. However, few substrates of GzmM have been reported to date, and the mechanism by
Robert van Domselaar et al.
Journal of immunology (Baltimore, Md. : 1950), 185(12), 7605-7613 (2010-11-10)
Granzyme M (GrM) is highly expressed in cytotoxic granules of NK cells, which provide the first line of defense against viral pathogens. GrM knockout mice show increased susceptibility toward murine CMV infection. Although GrM is a potent inducer of cell
Rukhshan Khurshid et al.
Molecular biology reports, 38(5), 2953-2960 (2010-01-29)
Granzymes kill cells in a variety of ways. They induce mitochondrial dysfunction through caspase dependent and caspase-independent pathways and destroy DNA and the integrity of the nucleus. For gaining a better understanding of the molecular function of granzyme M and
Pieter J A de Koning et al.
Molecular immunology, 47(4), 903-911 (2009-11-10)
The cytotoxic serine protease granzyme M (GrM) is one of the five human granzymes, which are mainly expressed by cytotoxic T lymphocytes and/or NK cells. Upon perforin-dependent entry into a target cell, GrM cleaves specific substrates resulting in the onset
Soo Jung Lee et al.
PloS one, 15(1), e0227672-e0227672 (2020-01-17)
A large number of pre-clinical and developmental investigations involve experimental vertebrate animals, of which mice have emerged as a favored organism. Recognition of the differences between humans and mice is essential for assessment of the relevance of animal studies to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service