Skip to Content
Merck
All Photos(4)

Key Documents

HPA012728

Sigma-Aldrich

Anti-VCP antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-5S Mg(2+)- ATPase p97 subunit, Anti-FANCG antibody produced in rabbit, Anti-TER ATPase, Anti-Transitional endoplasmic reticulum ATPase, Anti-VCP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

mouse, human, rat

enhanced validation

RNAi knockdown
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:1000-1:2500

immunogen sequence

LSDDTCSDEKIRMNRVVRNNLRVRLGDVISIQPCPDVKYGKRIHVLPIDDTVEGITGNLFEVYLKPYFLEAYRPIRKGDIFLVRGGMRAVEFKVVETDPSPYCIVAPDTVIHCEGEPIKREDEEESLNEVGYDDIGGCRKQLAQIKEMVE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FANCG(2189)

General description

Transitional endoplasmic reticulum ATPase (VCP) is an ATPase belonging to the ATPases associated with diverse cellular activities (AAA-ATPase) superfamily. The gene encoding VCP is located on chromosome 9pter-q34.

Immunogen

Transitional endoplasmic reticulum ATPase recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Transitional endoplasmic reticulum ATPase (VCP) binds to the C-terminal region of Dorfin (an ubiquitin ligase) and its ATPase activity contributes to the E3 activity of Dorfin. It helps glycoprotein 78 (Gp78) in coupling ubiquitination to degradation of 3-hydroxy-3-methylglutaryl coenzyme A reductase and has an important role in the recognition and degradation of endoplasmic reticulum-associated degradation (ERAD) substrates. VCP also aids ring finger protein 8 (RNF8) to mediate assembly of signaling complexes. Missense mutations in the VCP gene leads to inclusion body myopathy associated with Paget′s disease of bone and front temporal dementia. It is characterized by the presence of vacuoles in the cytoplasm of muscle fibers.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71607

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neeraj Vij
Journal of cellular and molecular medicine, 12(6A), 2511-2518 (2008-09-19)
p97/VCP, a member of the AAA-ATPase super family, has been associated with a wide variety of essential cellular protein pathways com prising: (i) nuclear envelope reconstruction, (ii) cell cycle, (iii) Golgi reassembly, (iv) suppression of apoptosis and (v) DNA-damage response
Giles D J Watts et al.
Nature genetics, 36(4), 377-381 (2004-03-23)
Inclusion body myopathy associated with Paget disease of bone and frontotemporal dementia (IBMPFD) is a dominant progressive disorder that maps to chromosome 9p21.1-p12. We investigated 13 families with IBMPFD linked to chromosome 9 using a candidate-gene approach. We found six
Shinsuke Ishigaki et al.
The Journal of biological chemistry, 279(49), 51376-51385 (2004-10-01)
Dorfin, a RING-IBR type ubiquitin ligase (E3), can ubiquitylate mutant superoxide dismutase 1, the causative gene of familial amyotrophic lateral sclerosis (ALS). Dorfin is located in ubiquitylated inclusions (UBIs) in various neurodegenerative disorders, such as ALS and Parkinson's disease (PD).
T Druck et al.
Genomics, 30(1), 94-97 (1995-11-01)
Clathrin-coated vesicles, involved in endocytosis and Golgi processing, have a surface lattice containing clathrin triskelia and stoichiometric amounts of additional components termed "assembly proteins," or APs. The AP form at the plasma membrane, AP2, is composed of two large subunits
Mayura Meerang et al.
Nature cell biology, 13(11), 1376-1382 (2011-10-25)
Unrepaired DNA double-strand breaks (DSBs) cause genetic instability that leads to malignant transformation or cell death. Cells respond to DSBs with the ordered recruitment of signalling and repair proteins to the site of lesion. Protein modification with ubiquitin is crucial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service