Skip to Content
Merck
All Photos(8)

Documents

HPA012123

Sigma-Aldrich

Anti-STIM1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Stromal interaction molecule 1 precursor

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

rat, human

enhanced validation

independent
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

VHPGSLVEKLPDSPALAKKALLALNHGLDKAHSLMELSPSAPPGGSPHLDSSRSHSPSSPDPDTPSPVGDSRALQASRNTRIPHLAGKKAVAEEDNGSIGEETDSSPGRKKFPLKIFK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... STIM1(6786)

Immunogen

Stromal interaction molecule 1 precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-STIM1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

Stromal Interaction Molecule 1 (STIM1) is a 90kDa cell surface transmembrane protein consisting of Ca2+ binding domains. It is localized in the endoplasmic reticulum (ER). In ER, depletion of stored Ca2+ ions activates the STIM1 and upon activation, it turns into multimeric form. These multimers translocate to the ER-plasma membrane junctions for the activation of Ca2+ channel. It mediates the store-operated calcium entry (SOCE) for normal cell functioning. It is also involved in the tumor growth suppression and stromal-haematopoietic cell interactions. Mutations in STIM1 cause autosomal-recessive hypomaturation enamel malformations.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72181

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Miriam Eckstein et al.
JCI insight, 2(6), e91166-e91166 (2017-03-30)
Loss-of-function mutations in stromal interaction molecule 1 (STIM1) impair the activation of Ca
Ga-Yeon Son et al.
Science signaling, 16(801), eadf9535-eadf9535 (2023-09-05)
Oral cancer causes pain associated with cancer progression. We report here that the function of the Ca2+ channel ORAI1 is an important regulator of oral cancer pain. ORAI1 was highly expressed in tumor samples from patients with oral cancer, and
Simon Latour et al.
Cancers, 10(11) (2018-10-31)
Ca2+ release-activated Ca2+ channels, composed of Orai1 and STIM1 (stromal interaction molecule 1) proteins, are the main Ca2+ entry mechanism in lymphocytes. Their role in cell migration and metastasis is demonstrated in solid cancers but it remains elusive in malignant
S Wang et al.
Journal of dental research, 93(7 Suppl), 94S-100S (2014-03-14)
Dental enamel formation depends upon the transcellular transport of Ca(2+) by ameloblasts, but little is known about the molecular mechanism, or even if the same process is operative during the secretory and maturation stages of amelogenesis. Identifying mutations in genes
S S Manji et al.
Biochimica et biophysica acta, 1481(1), 147-155 (2000-09-27)
STIM1 is a novel candidate growth suppressor gene mapping to the human chromosome region 11p15.5 that is associated with several malignancies. STIM1 overexpression studies in G401 rhabdoid tumour, rhabdomyosarcoma and rodent myoblast cell lines causes growth arrest, consistent with a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service