Skip to Content
Merck
All Photos(4)

Key Documents

HPA005633

Sigma-Aldrich

Anti-GATA2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Endothelial transcription factor GATA-2 antibody produced in rabbit, Anti-GATA-binding protein 2 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

LTGGQMCRPHLLHSPGLPWLDGGKAALSAAAAHHHNPWTVSPFSKTPLHPSAAGGPGGPLSVYPGAGGGSGGGSGSSVASLTPTAAHSGSHLFGFPPTPPKEVSPDPSTTGAASPASSSAGGSAARG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GATA2(2624)

General description

GATA-binding protein 2 (GATA2) is a transcription factor involved in several neuronal specification pathways and expressed in the adipose tissue. The gene is mapped to human chromosome 3q21.3. It encodes a zinc finger protein belonging to the GATA family of proteins.

Immunogen

Endothelial transcription factor GATA-2 recombinant protein epitope signature tag (PrEST)

Application

Anti-GATA2 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige. Anti-GATA2 antibody produced in rabbit has been used in immunoprecipitation.

Biochem/physiol Actions

GATA-binding protein 2 (GATA2) blocks the transition from preadipocytes to adipocytes through suppression of PPAR-γ2 expression. GATA2 is also known to direct hematopoiesis and vascular cell activation. Thus, preadipocytes inhibition of GATA2 could allow adipogenesis while reducing their inflammatory properties. It is an inhibitor of adipogenesis and an activator of vascular cells. GATA2 is phosphorylated and blocked by the PI3K/Akt signal transduction pathway, this phosphorylation results in preadipocyte conversion to adipocytes and attenuation of the inflammatory behavior of preadipocytes. The encoded protein is essential for the development of CNS and the differentaition of erythroid cells. Polymorphism in this gene has been associated with Parkinson′s disease.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST83056

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Harriet Holme et al.
British journal of haematology, 158(2), 242-248 (2012-04-27)
The myelodysplastic syndromes (MDS) are heterogeneous and can evolve into acute myeloid leukaemia (AML). Rare familial cases are reported in which five disease genes have been identified to date (RUNX1, CEBPA, TERC, TERT and GATA2). Here we report the genetic
Amanda Kohlmeier et al.
Endocrinology, 161(6) (2020-04-27)
The transcription factor GATA2 is important for endometrial stromal cell decidualization in early pregnancy. Progesterone receptor (PGR) is also critical during decidualization but its interaction with GATA2 in regulating genes and pathways necessary for decidualization in human endometrium are unclear.
Lewis Chaytor et al.
Molecular cancer research : MCR, 17(6), 1264-1278 (2019-03-06)
The androgen receptor (AR) is a key driver of prostate cancer development. Antiandrogens effectively inactivate the AR, but subsequent AR reactivation progresses the disease to castrate-resistant prostate cancer (CRPC). Constitutively active AR splice variants (AR-V) that function unchallenged by current
Abeer El Wakil et al.
Development (Cambridge, England), 133(11), 2155-2165 (2006-05-05)
Postmitotic neurons are produced from a pool of cycling progenitors in an orderly fashion that requires proper spatial and temporal coordination of proliferation, fate determination, differentiation and morphogenesis. This probably relies on complex interplay between mechanisms that control cell cycle
Veronica Rodriguez-Bravo et al.
Cell, 174(5), 1200-1215 (2018-08-14)
Nuclear pore complexes (NPCs) regulate nuclear-cytoplasmic transport, transcription, and genome integrity in eukaryotic cells. However, their functional roles in cancer remain poorly understood. We interrogated the evolutionary transcriptomic landscape of NPC components, nucleoporins (Nups), from primary to advanced metastatic human

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service