Skip to Content
Merck
All Photos(3)

Key Documents

HPA004177

Sigma-Aldrich

Anti-ALDOA antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Fructose-bisphosphate aldolase A antibody produced in rabbit, Anti-Lung cancer antigen NY-LU-1 antibody produced in rabbit, Anti-Muscle-type aldolase antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

IVAPGKGILAADESTGSIAKRLQSIGTENTEENRRFYRQLLLTADDRVNPCIGGVILFHETLYQKADDGRPFPQVIKSKGGVVGIKVDKGVVPLAGTNGETTTQGL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ALDOA(226)

General description

Aldolase A, fructose-bisphosphate (ALDOA) is a ubiquitous glycolytic enzyme expressed in developing embryo and in adult muscle. It is involved in a wide range of cellular functions such as maintenance of striated muscle contraction, cell shape and mobility, actin filament organization and ATP biosynthetic process.

Immunogen

Fructose-bisphosphate aldolase A recombinant protein epitope signature tag (PrEST)

Application

Anti-ALDOA antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

In glycolysis, ALDOA catalyzes the reversible reaction of fructose-1,6-bisphosphate to glyceraldehydes-3-phosphate and dihydroxyacetone phosphate. High expression level of ALDOA is reported in various forms of malignant cancers, including human lung squamous, renal cell and hepatocellular carcinomas. ALDOA deficiency causes myopathy and hemolytic anemia.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST85927

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sarah Morson et al.
Cerebral cortex (New York, N.Y. : 1991), 31(9), 4038-4052 (2021-04-08)
The 593 kbp 16p11.2 copy number variation (CNV) affects the gene dosage of 29 protein coding genes, with heterozygous 16p11.2 microduplication or microdeletion implicated in about 1% of autism spectrum disorder (ASD) cases. The 16p11.2 CNV is frequently associated with
Qiwen Kuang et al.
OncoTargets and therapy, 14, 3353-3366 (2021-06-04)
ALDOA plays an essential role in cancer progression in different human cancers; however, its function has not been understood in prostate cancer (PCa). Associations of ALDOA expression with clinicopathological features and patient prognosis in PCa were evaluated based on data
Ning Na et al.
Therapeutics and clinical risk management, 13, 279-285 (2017-03-11)
Aldolase A (ALDOA) is a glycolytic enzyme that drives the glycolytic metabolic pathway in mammalian cells. The overexpression of ALDOA was observed in a variety of cancers including clear-cell renal cell carcinoma (ccRCC). However, little was known about the clinicopathological
S Miwa et al.
American journal of hematology, 11(4), 425-437 (1981-12-01)
Two cases of red cell aldolase deficiency associated with congenital nonspherocytic hemolytic anemia are reported. The proband is a fourteen-month-old Japanese boy. Consanguineous marriage was not proven but probable in this family, as the parents were born in the same
Seyedehmahsa Moghimi et al.
PLoS pathogens, 18(10), e1010906-e1010906 (2022-10-29)
As ultimate parasites, viruses depend on host factors for every step of their life cycle. On the other hand, cells evolved multiple mechanisms of detecting and interfering with viral replication. Yet, our understanding of the complex ensembles of pro- and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service