Skip to Content
Merck
All Photos(5)

Key Documents

HPA004135

Sigma-Aldrich

Anti-CRABP2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CRABP-II, Anti-Cellular retinoic acid-binding protein 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

IKQEGDTFYIKTSTTVRTTEINFKVGEEFEEQTVDGRPCKSLVKWESENKMVCEQKLLKGEGPKTSWTRELTNDGELILTMTADDVVCTRVYVRE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CRABP2(1382)

General description

Cellular retinoic acid binding protein 2 (CRABP2) is an intracellular lipid-binding protein, involved in the stimulation of receptor transcriptional activity. Reports show that, CRABP-II contains three potential SUMOylation sites centered at K45, K87, and K102.

Immunogen

Cellular retinoic acid-binding protein 2 recombinant protein epitope signature tag (PrEST)

Application

Anti-CRABP2 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

Cellular retinoic acid binding protein 2 (CRABP2) plays a crucial role in acceleration of the transcriptional activity. In the nucleus, by binding directly to the nuclear receptor RAR, it forms a complex for connecting retinoic acid (RA) from the binding protein to the receptor. This interaction finally helps to accelerate the transcriptional activity. It may have impact in the immature phenotype in glioma cells.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST79911

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Irina Favorskaya et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 35(10), 10295-10300 (2014-07-19)
The impairment of retinoic acid (RA)-dependent signaling is a frequent event during carcinogenesis. Cellular retinoic acid-binding proteins (CRABP1 and CRABP2) are important modulators of RA activity. Up to date, the role of these proteins in cancer progression remains poorly investigated.
Avijit Majumdar et al.
The Journal of biological chemistry, 286(49), 42749-42757 (2011-10-15)
Cellular retinoic acid-binding protein II (CRABP-II) undergoes nuclear translocation upon binding of retinoic acid (RA). In the nucleus, CRABP-II directly binds to the nuclear receptor RAR to form a complex through which RA is "channeled" from the binding protein to
Benito Campos et al.
International journal of cancer, 131(8), 1963-1968 (2012-01-26)
Impairment of endogenous differentiation pathways like retinoic acid (RA) signaling seems to be a central pathogenetic event in astrocytic gliomas. Among others, expression of the differentiation-promoting RA chaperon protein cellular retinoic acid binding protein 2 (CRABP2) is extenuated in high-grade
Rie Ozaki et al.
PloS one, 12(3), e0173035-e0173035 (2017-03-03)
Upon breaching of the endometrial surface epithelium, the implanting embryo embeds in the decidualizing stroma. Retinoic acid (RA), a metabolite of vitamin A, is an important morphogen during embryonic and fetal development, although the role of the RA pathway in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service