Skip to Content
Merck
All Photos(7)

Key Documents

HPA003595

Sigma-Aldrich

Anti-ARG1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Arginase-1 antibody produced in rabbit, Anti-Liver-type arginase antibody produced in rabbit, Anti-Type I arginase antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:2500-1:5000

immunogen sequence

TIGIIGAPFSKGQPRGGVEEGPTVLRKAGLLEKLKEQECDVKDYGDLPFADIPNDSPFQIVKNPRSVGKASEQLAGKVAEVKKNGRISLVLGGDHSLAIGSISGHARVHPDLGVIWVDA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ARG1(383)

Looking for similar products? Visit Product Comparison Guide

General description

ARG1 (arginase 1) is an enzyme responsible for the processing of the essential amino acid arginine, to produce urea and L-ornithine. This enzyme is expressed in the presence of Th2 (T-helper) cytokines such as, interleukin (IL-4), IL-13 and TGFβ (tumor growth factor).

Immunogen

Arginase-1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

The human ARG1 gene encodes the protein arginase. During trauma, cancer, chronic wounds, pregnancy and diabetes the serum arginase levels is found to be elevated. It shows superior sensitivity and specificity in the diagnosis of HCC (Hepatocellular Carcinoma) while it has low sensitivity and a very high specificity for pancreatic adenocarcinoma (PAD). It might regulate serum L-arginine and 3-nitrotyrosine through L-arginine. Hepatocyte arginase deficiency causes argininemia (an autosomal recessive genetic disorder).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST83046

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mika Fujiwara et al.
Cancer cytopathology, 120(4), 230-237 (2012-03-22)
Distinguishing hepatocellular carcinoma (HCC) from adenocarcinoma in fine-needle aspiration biopsies (FNAB) is often diagnostically challenging. Arginase-1 was recently described as a marker of hepatic differentiation in surgical resection specimens. We compared the expression of arginase-1, HepPar-1, and glypican-3 in FNAB
Congrong Wang et al.
Translational cancer research, 9(1), 128-136 (2020-01-01)
The aim of the present study was to investigate the diagnostic value of glypican-3 (GPC3), arginase-1 (Arg-1), and hepatocyte paraffin antigen 1 (HepPar-1) in differentiating hepatocellular carcinoma (HCC) from intrahepatic cholangiocarcinoma (ICC). The expression of GPC3, HepPar-1 and Arg-1 were
Andrés Vacas et al.
Biomedicines, 8(11) (2020-10-30)
A novel serine/threonine protein kinase, LmjF.22.0810, was recently described in Leishmania major. After generating an L. major cell line overexpressing LmjF.22.0810 (named LmJ3OE), the ability of this novel protein to modulate the Th2-type immune response was analyzed. Our results suggest
Juana Schwartz et al.
Journal of dermatological science, 92(1), 78-88 (2018-07-25)
Cutaneous leishmaniasis (CL) skin lesions are the result of a deregulated immune response, which is unable to eliminate Leishmania parasites. The control of both, parasites and host immune response, is critical to prevent tissue destruction. The skin ulceration has been
K Ogino et al.
Free radical research, 48(2), 137-145 (2013-09-26)
The associations of serum arginase I with serum L-arginine, serum 3-nitrotyrosine, and fractional exhaled nitric oxide (FENO) were evaluated cross-sectionally in healthy Japanese workers. The serum median (minimum-maximum) levels of arginase I, 3-nitrotyrosine, and FENO in healthy people (n =

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service