Skip to Content
Merck
All Photos(7)

Key Documents

HPA001352

Sigma-Aldrich

Anti-APOA4 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Apo-AIV antibody produced in rabbit, Anti-ApoA-IV antibody produced in rabbit, Anti-Apolipoprotein A-IV precursor antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:500-1:1000
western blot: 0.04-0.4 μg/mL

immunogen sequence

LEGLTFQMKKNAEELKARISASAEELRQRLAPLAEDVRGNLRGNTEGLQKSLAELGGHLDQQVEEFRRRVEPYGENFNKALVQQMEQLRQKLGPHAGDVEGHLSFLEKDLRDKVNSFFSTFKEKESQDKTLSLP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... APOA4(337)

Looking for similar products? Visit Product Comparison Guide

General description

The gene APOA4 is localized to the long arm of human chromosome 11 along with A1 (APOA1), C3 (APOC3) genes. This gene contains three exons separated by two introns. Apo A-IV is synthesized in the mucosal cells of the small intestine as a preprotein. This 396-amino acid preprotein undergoes proteolytic processing, associates with chylomicrons during fat absorption and is secreted into the lymph.

Immunogen

Apolipoprotein A-IV precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

APOA4 (apolipoprotein A-IV) gene encodes a 396-amino acid protein that forms a component of the lipid transport system. It is found in association with chylomicrons, high-density lipoprotein (HDL), and the lipoprotein-free fraction of the plasma. It is involved in the metabolism of high density lipoproteins and chylomicrons. Human apo A-IV is found to be a potent activator of the enzyme lecithin-cholesterol acyltransferase in vitro.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST77456

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Steinmetz et al.
The Journal of biological chemistry, 260(4), 2258-2264 (1985-02-25)
Human plasma apoproteins (apo) A-I and A-IV both activate the enzyme lecithin:cholesterol acyltransferase (EC 2.3.1.43). Lecithin:cholesterol acyltransferase activity was measured by the conversion of [4-14C] cholesterol to [4-14C]cholesteryl ester using artificial phospholipid/cholesterol/[4-14C]cholesterol/apoprotein substrates. The substrate was prepared by the addition
S K Karathanasis et al.
Biochemistry, 25(13), 3962-3970 (1986-07-01)
Apolipoprotein AIV (apoAIV) is a protein of the lipid transport system found associated with chylomicrons, high-density lipoprotein (HDL), and the lipoprotein-free fraction of the plasma. The gene coding for the human apoAIV is closely linked with the genes coding for
H Tenkanen et al.
Arteriosclerosis and thrombosis : a journal of vascular biology, 11(4), 851-856 (1991-07-01)
Apolipoprotein (apo) A-IV is a protein involved in the metabolism of chylomicrons and high density lipoproteins. This protein displays genetic polymorphism due to two main codominant alleles, A-IV1 and A-IV2. We have identified the mutation that leads to this polymorphism.
Hiten D Mistry et al.
Journal of lipid research, 58(6), 1186-1195 (2017-04-12)
Preeclampsia is a pregnancy-specific condition that leads to increased cardiovascular risk in later life. A decrease in cholesterol efflux capacity is linked to CVD. We hypothesized that in preeclampsia there would be a disruption of maternal/fetal plasma to efflux cholesterol
N A Elshourbagy et al.
The Journal of biological chemistry, 262(17), 7973-7981 (1987-06-15)
We have isolated the human apolipoprotein (apo) A-IV gene from a cosmid library and determined its complete nucleotide sequence. The gene contains three exons of 162, 127, and 1180 nucleotides separated by two introns of 357 and 777 nucleotides. A

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service